Skip to Content
Merck
All Photos(1)

Documents

M3189

Sigma-Aldrich

Mepivacaine hydrochloride

98.0-102.0%, meets USP testing specifications

Synonym(s):

1-Methyl-2′,6′-pipecoloxylidine hydrochloride, N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O · HCl
CAS Number:
Molecular Weight:
282.81
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

meets USP testing specifications

Assay

98.0-102.0%

originator

Novocol

SMILES string

CN1CCCCC1C(NC2=C(C)C=CC=C2C)=O.Cl

InChI

1S/C15H22N2O.ClH/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3;/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18);1H

InChI key

RETIMRUQNCDCQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Local anesthetic. Reversibly blocks transient Na+ inward current, as well as the steady-state K+ outward current. Blocks tandem pore (TASK) and Kv1.5, potassium channels in model systems.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novocol. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Van Vynckt et al.
Veterinary and comparative orthopaedics and traumatology : V.C.O.T, 25(4), 307-313 (2012-06-15)
Assessment of the effect of intra-articular anaesthesia on lameness caused by medial coronoid disease. This study included 90 dogs that were evaluated for the complaint of unilateral forelimb lameness. All dogs were suspected of having an elbow problem for which
Hui-Jin Sung et al.
Canadian journal of physiology and pharmacology, 90(7), 863-872 (2012-06-19)
Mepivacaine is an aminoamide-linked local anesthetic with an intermediate duration that intrinsically produces vasoconstriction both in vivo and in vitro. The aims of this in-vitro study were to examine the direct effect of mepivacaine in isolated rat aortic rings and
Mi Hyeon Lee et al.
Pain medicine (Malden, Mass.), 13(11), 1381-1388 (2012-09-28)
Compared with the blind technique, ultrasound-guided stellate ganglion block (SGB) reduces the amount of local anesthetic needed for a successful block. The purpose of this study is to determine the minimal, optimal volume of local anesthetic required for successful ultrasound-guided
Jeff Gadsden et al.
Anesthesia and analgesia, 115(4), 963-967 (2012-07-17)
During peripheral nerve blockade, different local anesthetics may be sequentially administered. Typically, a short- or intermediate-acting local anesthetic is administered before a long-acting local anesthetic to achieve a block with rapid onset and long duration. However, there is a paucity
Mylène Lecours et al.
Canadian journal of anaesthesia = Journal canadien d'anesthesie, 60(3), 244-252 (2012-12-06)
In recent studies on ultrasound-guided infraclavicular block (ICB), the authors have favoured a single injection posterior to the axillary artery rather than multiple injections; however, procedural complications and success rates associated with single-injection ultrasound-guided ICB are not well known. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service