Skip to Content
Merck
All Photos(1)

Key Documents

M3064

Sigma-Aldrich

Myostatin human

recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

GDF-8, Growth Differentiation Factor - 8

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
R$8,948.00

R$8,948.00


Check Cart for Availability


Select a Size

Change View
0.1 MG
R$8,948.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

R$8,948.00


Check Cart for Availability

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

16 kDa (including a 31 amino-acid spacer and 6-His tag)

packaging

pkg of 0.1 mg

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... MSTN(2660)

Application

Use recombinant myostatin to study its mechanisms of action in vitro.

Biochem/physiol Actions

Myostatin is a 25 KDa, homodimer, that acts on muscle to induce SMAD family transcription factors such as SMAD2 and SMAD3. It inhibits the differentiation of myoblasts into mature muscle fibers. Positive control for myostatin in Western blots and ELISA.
Positive control for myostatin in Western blots and ELISA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Takahashi et al.
Domestic animal endocrinology, 48, 62-68 (2014-06-08)
The purpose of this study was to determine whether myostatin alters glucose transporter-4 (GLUT4) expression in bovine skeletal muscles and myoblasts isolated from double-muscled (DM) and normal-muscled (NM) Japanese Shorthorn cattle. Plasma concentrations of glucose were lower in DM cattle
Karima Relizani et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 22(8), 1423-1433 (2014-05-28)
Myostatin regulates skeletal muscle size via the activin receptor IIB (ActRIIB). However, its effect on muscle energy metabolism and energy-dependent muscle function remains largely unexplored. This question needs to be solved urgently since various therapies for neuromuscular diseases based on
R E Ferrell et al.
Genomics, 62(2), 203-207 (1999-12-28)
Myostatin is a recently identified member of the transforming growth factor-beta family of regulatory factors, also known as growth and differentiation factor 8 (GDF8). The nucleotide sequence of human myostatin was determined in 40 individuals. The invariant promoter contains a
Qiang Zhang et al.
The FEBS journal, 277(2), 466-476 (2009-12-18)
The ubiquitin ligase RING finger protein 13 gene (RNF13) was first identified in a screen for genes whose expression is regulated by myostatin in chicken fetal myoblasts. In this study, we demonstrate that the RNF13 gene is broadly expressed in
Yutaka Ohsawa et al.
PloS one, 10(7), e0133713-e0133713 (2015-08-01)
Myostatin, a muscle-specific transforming growth factor-β (TGF-β), negatively regulates skeletal muscle mass. The N-terminal prodomain of myostatin noncovalently binds to and suppresses the C-terminal mature domain (ligand) as an inactive circulating complex. However, which region of the myostatin prodomain is

Questions

  1. Is this the active/mature myostatin protein or does is it the proprotein before being activated?

    1 answer
    1. This product is not an active/mature myostatin protein. The sequence used for this myostatin protein is residues 267 - 375. The recombinant protein also includes an N-terminal His tag followed by a spacer before the myostatin sequence.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service