Skip to Content
Merck
All Photos(1)

Key Documents

M2776

Sigma-Aldrich

Methylergonovine maleate salt

≥98% (TLC)

Synonym(s):

9,10-Didehydro-N-[(S)-1-(hydroxymethyl)propyl]-6-methylergoline-8β-carboxamide maleate salt, Methergine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N3O2 · C4H4O4
CAS Number:
Molecular Weight:
455.50
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

color

white

solubility

H2O: 25 mg/mL
0.1 M HCl: soluble
ethanol: slightly soluble

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.[H][C@@]12Cc3c[nH]c4cccc(C1=C[C@H](CN2C)C(=O)N[C@@H](CC)CO)c34

InChI

1S/C20H25N3O2.C4H4O4/c1-3-14(11-24)22-20(25)13-7-16-15-5-4-6-17-19(15)12(9-21-17)8-18(16)23(2)10-13;5-3(6)1-2-4(7)8/h4-7,9,13-14,18,21,24H,3,8,10-11H2,1-2H3,(H,22,25);1-2H,(H,5,6)(H,7,8)/b;2-1-/t13-,14+,18-;/m1./s1

InChI key

NOFOWWRHEPHDCY-DAUURJMHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dopamine antagonist interacts with serotonergic, dopaminergic and α-adrenergic systems.
Ergot alkaloid which interacts with serotonergic, dopaminergic and α-adrenergic systems. Dopamine antagonist.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axel de Labriolle et al.
Cardiovascular toxicology, 9(1), 46-48 (2009-02-17)
Ergot derivatives are frequently administrated during cesarean delivery, induced abortion, or post-partum hemorrhage to promote uterine contractions. Ergot derivatives may also induce coronary spasm and intravenous ergonovine is used in cardiac catheterization laboratories as a diagnostic agent. Serious ischemic cardiac
C C Enweronu-Laryea et al.
East African medical journal, 85(9), 463-465 (2009-06-23)
Methylergometrine is probably the most commonly used drug in obstetric care at all levels of the health care system. Many communities in Africa lack skilled birth attendants and adequate health systems; medication errors are more likely to occur and go
Kazutoshi Isobe et al.
Nihon Kokyuki Gakkai zasshi = the journal of the Japanese Respiratory Society, 46(12), 1007-1012 (2009-02-07)
A 35-year-old woman underwent endometrial curettage for suspicion of miscarriage. A few minutes after intravenous injection of methylergometrin (0.2 mg) for inducing uterine contraction, blood gas analysis revealed severe hypoxemia. Chest CT showed diffuse ground-glass opacities in both lung fields
Falah Aboukhoudir et al.
European journal of echocardiography : the journal of the Working Group on Echocardiography of the European Society of Cardiology, 10(4), 556-561 (2009-02-10)
The aim of this article was to assess whether abnormal dobutamine stress echocardiography (DSE) can be due to a dobutamine-induced coronary spasm in patients with angiographically documented vasospastic coronary arteries. Between January 2004 and April 2008, we prospectively evaluated all
Sudden death caused by atypical variant angina.
Franck Laporte et al.
Archives of cardiovascular diseases, 104(8-9), 480-481 (2011-09-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service