Skip to Content
Merck
All Photos(1)

Key Documents

M0824

Sigma-Aldrich

MSSK1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC102944, SRPK3, STK23

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$4,292.00

R$4,292.00


Estimated to ship onMay 22, 2025



Select a Size

Change View
10 μG
R$4,292.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

R$4,292.00


Estimated to ship onMay 22, 2025


recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

200-270 nmol/min·mg

mol wt

~98 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SRPK3(26576)

Biochem/physiol Actions

MSSK1, also known as SRPK3, is a muscle-specific protein kinase belonging to the serine arginine protein kinase family which phosphorylates serine/arginine repeat-containing proteins. Heart and skeletal muscle show high expression of the MSSK1/SRPK3 gene product. SRPK3-null mice display a new entity of type 2 fiber-specific myopathy with a marked increase in centrally placed nuclei. Transgenic mice overexpressing SRPK3 in skeletal muscle show severe myofiber degeneration and early lethality.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osamu Nakagawa et al.
Genes & development, 19(17), 2066-2077 (2005-09-06)
Myocyte enhancer factor 2 (MEF2) plays essential roles in transcriptional control of muscle development. However, signaling pathways acting downstream of MEF2 are largely unknown. Here, we performed a microarray analysis using Mef2c-null mouse embryos and identified a novel MEF2-regulated gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service