Skip to Content
Merck
All Photos(1)

Key Documents

M053030

Sigma-Aldrich

DMT-5-Methyl-dC(ac) Phosphoramidite

configured for ABI

Synonym(s):

5-Methyl-dC(ac) amidite

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
R$1,120.00

R$1,120.00


Check Cart for Availability


Select a Size

Change View
0.25 G
R$1,120.00

About This Item

Empirical Formula (Hill Notation):
C42H52N5O8P
Molecular Weight:
785.86
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

R$1,120.00


Check Cart for Availability

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

λ

conforms (UV/VIS Identity)

compatibility

configured for ABI

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=C(C)C(NC(C)=O)=NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C42H52N5O8P/c1-28(2)47(29(3)4)56(53-24-12-23-43)55-37-25-39(46-26-30(5)40(44-31(6)48)45-41(46)49)54-38(37)27-52-42(32-13-10-9-11-14-32,33-15-19-35(50-7)20-16-33)34-17-21-36(51-8)22-18-34/h9-11,13-22,26,28-29,37-39H,12,24-25,27H2,1-8H3,(H,44,45,48,49)/t37-,38+,39+,56?/m0/s1

InChI key

YFUQEQDYBKCNTE-CMFYRTQUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service