Skip to Content
Merck
All Photos(1)

Key Documents

M021030

Sigma-Aldrich

MMT-Hexylaminolinker Phosphoramidite

configured for ABI

Synonym(s):

MMT-Hexylamine-linker amidite

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
R$866.00

R$866.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.25 G
R$866.00

About This Item

Empirical Formula (Hill Notation):
C35H48N3O3P
CAS Number:
Molecular Weight:
589.75
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

R$866.00


Check Cart for Availability

Request a Bulk Order

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

Assay

≥97% (reversed phase HPLC)
≥98% (31P-NMR)

suitability

conforms to structure for H-NMR

compatibility

configured for ABI

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(NCCCCCCOP(OCCC#N)N(C(C)C)C(C)C)(c2ccccc2)c3ccccc3

InChI

1S/C35H48N3O3P/c1-29(2)38(30(3)4)42(41-28-16-25-36)40-27-15-7-6-14-26-37-35(31-17-10-8-11-18-31,32-19-12-9-13-20-32)33-21-23-34(39-5)24-22-33/h8-13,17-24,29-30,37H,6-7,14-16,26-28H2,1-5H3

InChI key

YBANXOPIYSVPMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service