Skip to Content
Merck
All Photos(1)

Documents

L6788

Sigma-Aldrich

Leptin Fragment 116-130 Amide mouse

>95% (HPLC), white, powder

Synonym(s):

Obese Gene Peptide 116-130 Amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H109N19O24S
CAS Number:
Molecular Weight:
1560.73
MDL number:
UNSPSC Code:
51111800

Assay

>95% (HPLC)

form

powder

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Lep(16846)

Amino Acid Sequence

Ser-Cys-Ser-Leu-Pro-Gln-Thr-Ser-Gly-Leu-Gln-Lys-Pro-Glu-Ser-NH2

Biochem/physiol Actions

Mimics the effects of recombinant leptin on body weight and food intake.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service