Skip to Content
Merck
All Photos(1)

Key Documents

L6670

Sigma-Aldrich

LY266097

≥98% (HPLC), powder

Synonym(s):

1-[(2-Chloro-3,4-dimethoxyphenyl)methyl]-2,3,4,9-tetrahydro-6-methyl-1H-pyrido[3,4-b]indole hydrochloride, LY 266097

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClN2O2 · HCl
CAS Number:
Molecular Weight:
407.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥20 mg/mL

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(CC2NCCc3c2[nH]c4ccc(C)cc34)c(Cl)c1OC

InChI

1S/C21H23ClN2O2.ClH/c1-12-4-6-16-15(10-12)14-8-9-23-17(20(14)24-16)11-13-5-7-18(25-2)21(26-3)19(13)22;/h4-7,10,17,23-24H,8-9,11H2,1-3H3;1H

InChI key

KPXKZZURYAXZQE-UHFFFAOYSA-N

Biochem/physiol Actions

LY266097 is a selective 5HT-2B antagonist with a pKI of 9.7 for the human cloned 5-HT2B receptor and a 100-fold greater selectivity for 5-HT2B than the human 5-HT2C and 5-HT2A.

Features and Benefits

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Céline Devroye et al.
Neuropharmacology, 109, 59-68 (2016-06-05)
Recent studies suggest that the central serotonin2B receptor (5-HT2BR) could be an interesting pharmacological target for treating neuropsychiatric disorders related to dopamine (DA) dysfunction, such as schizophrenia. Thus, the present study was aimed at characterizing the role of 5-HT2BRs in

Articles

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service