Skip to Content
Merck
All Photos(1)

Documents

L6133

Sigma-Aldrich

Lichenan from Cetraria islandica

practical grade, powder

Synonym(s):

Lichenin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

practical grade

form

powder

color

tan to brown

SMILES string

OC[C@@H]1O[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O

Looking for similar products? Visit Product Comparison Guide

Other Notes

A glucose polysaccharide.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service