Skip to Content
Merck
All Photos(1)

Key Documents

L137

Sigma-Aldrich

L-733,060 hydrochloride

solid, ≥98% (HPLC)

Synonym(s):

(2S,3S)-3-[(3,5-bis(Trifluoromethyl)phenyl)methoxy]-2-phenylpiperidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19F6NO · HCl
CAS Number:
Molecular Weight:
439.82
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Cl[H].FC(F)(F)c1cc(CO[C@@H]2CCCN[C@@H]2c3ccccc3)cc(c1)C(F)(F)F

Gene Information

human ... TACR1(6869)

Biochem/physiol Actions

Non-peptide NK1 tachykinin receptor antagonist; selective for guinea pig/human versus rat/mouse. Micromolar concentrations of L-733,060 inhibit the growth of human gastric and colon adenocarcinomas.in a dose-dependent manner, eventually leading to apoptosis in both cell lines.

Legal Information

Manufactured and sold under license from Merck & Co., Inc., Kenilworth, NJ, U.S.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Kiatpapan et al.
Journal of bioscience and bioengineering, 92(5), 459-465 (2005-10-20)
To develop systems for the expression of heterologous genes in probiotic strains of Lactobacillus and Propionibacterium, we used Lactobacillus plantarum and Propionibacterium freudenreichii and a modified gene encoding cholesterol oxidase (choA) from Streptomyces sp. to generate working models. The acetyl
Jitka Myskova et al.
Journal of medical entomology, 45(1), 133-138 (2008-02-21)
Quantification of Leishmania parasites in the sand fly digestive tract is important for evaluation of vector competence. We compared quantitative polymerase chain reaction (Q-PCR) with two "traditional" methods, estimation in situ and direct counting with the aid of a hemocytometer
Jong-Hyun Kim et al.
Journal of bioscience and bioengineering, 107(2), 124-129 (2009-02-17)
A gene (apuA) encoding amylopullulanase from a starch-hydrolyzing lactic acid bacterium, Lactobacillus plantarum L137, which had been isolated from traditional fermented food made from fish and rice in the Philippines, was found to contain two unique amino acid repeating units
Y Kaneko et al.
Journal of bioscience and bioengineering, 89(1), 62-67 (2005-10-20)
Lactobacillus plantarum NC13, a strain derived from the L. plantarum strain L137 isolated from a traditional fermented food produced in the Philippines, contains 9 of the 15 plasmids in the parental strain. To construct a shuttle vector between L. plantarum
N M Rupniak et al.
Pain, 67(1), 189-195 (1996-09-01)
Intravenous administration of the NK1 receptor antagonist L-733,060 to gerbils 3 h before intraplantar injection of formalin caused a dose-dependent and complete inhibition of the late, but not early, phase nociceptive response (paw licking). The ID50 for L-733,060 (0.17 mg/kg)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service