Skip to Content
Merck
All Photos(1)

Documents

L1277

Sigma-Aldrich

Lectin from Lens culinaris (lentil)

Isoelectric focusing marker, pI (1) 8.2, (2) 8.6, (3) 8.8

Synonym(s):

Lens culinaris agglutinin, LcH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

packaging

vial of ~1 mg protein

pI 

(1) 8.2, (2) 8.6, (3) 8.8

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

LcH is not blood group specific. It has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. LcH is comprised of two isomers, LcH-A (mol. wt. 49,000) and LcH-B (mol. wt. 49,000). LcH-A is mitogenic. Conjugates are prepared from purified lectin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taha Azad et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 29(6), 1984-2000 (2021-02-13)
The ongoing COVID-19 pandemic has highlighted the immediate need for the development of antiviral therapeutics targeting different stages of the SARS-CoV-2 life cycle. We developed a bioluminescence-based bioreporter to interrogate the interaction between the SARS-CoV-2 viral spike (S) protein and

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service