Skip to Content
Merck
All Photos(1)

Key Documents

L0665

Sigma-Aldrich

Lambda Light Chains (Free) from human myeloma sera

purified immunoglobulin, ~1.0 mg/mL, ≥99% (Purity by double diffusion), buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$4,592.00

R$4,592.00


Check Cart for Availability


Select a Size

Change View
1 MG
R$4,592.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

R$4,592.00


Check Cart for Availability

biological source

human

Quality Level

antibody form

purified immunoglobulin

Assay

≥99% (Purity by double diffusion)

form

buffered aqueous solution

concentration

~1.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulins (Ig) are produced by the B cells and play crucial role in host humoral immune response. A typical immunoglobulin consists of two heavy chains and two light chains. The two light chains are either κ or the λ chain. The B cells express one type of light chain during their life time[1]
λ Light Chains (Free) from human myeloma sera consists of IgG λ light chains purified from myeloma sera.

Application

λ Light Chains (Free) from human myeloma sera may be used as immunoglobulin calibrator, blocking agent or coating protein in various immunoassays.

Physical form

Solution in 0.01 M phosphate and 0.15 M sodium chloride, pH 7.2, containing 1.0 M glycine and 0.1% sodium azide.

Preparation Note

Purified from pooled IgG lambda myeloma sera.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerry A Katzmann et al.
Clinical chemistry, 48(9), 1437-1444 (2002-08-27)
The detection of monoclonal free light chains (FLCs) is an important diagnostic aid for a variety of monoclonal gammopathies and is especially important in light-chain diseases, such as light-chain myeloma, primary systemic amyloidosis, and light-chain-deposition disease. These diseases are more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service