Skip to Content
Merck
All Photos(1)

Key Documents

K4128

Sigma-Aldrich

Knudson C orchid medium Morel modification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.72
Pricing and availability is not currently available.

storage temp.

2-8°C

Quality Level

Formula variant

With the macro- and micronutrients as described by Morel (1965). Contains sucrose.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service