Skip to Content
Merck
All Photos(1)

Key Documents

K3518

Sigma-Aldrich

PIM2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
R$3,042.00

R$3,042.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
R$3,042.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

R$3,042.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

259-351 nmol/min·mg

mol wt

~61 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIM2(11040)

Application

PIM2, active, GST tagged human has been used for in vitro kinase assay in study of PIM kinases role in fms-like tyrosine kinase 3 inhibitors (FLT3i)-resistant acute myeloid leukemia.[1]

Biochem/physiol Actions

PIM2 (proto-oncogene) is a serine threonine kinase that is present in all tissues, being most abundant in hematopoietic tissues, testis, small instestine and colon, as well as in spleen and thymus. It is highly expressed in human leukemic, lymphoma and colorectal adenocarcinoma cell lines. This suggests a role for PIM2 in proliferating cells and during meiosis.[2] PIM2 acts as a pro-survival kinase and BAD (BCL2 associated agonist of cell death) protein is a PIM2 substrate.[3]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Baytel et al.
Biochimica et biophysica acta, 1442(2-3), 274-285 (1998-11-07)
In this study we describe the cloning of a human gene, encoding a protein that shares 90% identity and 93% similarity at the primary structure level, with the mouse Pim-2 gene. The gene was designated hPim-2. Structural features suggest that
Alexa S Green et al.
Science advances, 1(8), e1500221-e1500221 (2015-11-26)
Fms-like tyrosine kinase 3 internal tandem duplication (FLT3-ITD) is frequently detected in acute myeloid leukemia (AML) patients and is associated with a dismal long-term prognosis. FLT3 tyrosine kinase inhibitors provide short-term disease control, but relapse invariably occurs within months. Pim
Bin Yan et al.
The Journal of biological chemistry, 278(46), 45358-45367 (2003-09-05)
Hematopoietic growth factors mediate the survival and proliferation of blood-forming cells, but the mechanisms through which these proteins produce their effects are incompletely known. Recent studies have identified the pim family of kinases as mediators of cytokine-dependent survival signals. Several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service