Skip to Content
Merck
All Photos(1)

Key Documents

K1515

Sigma-Aldrich

Anti-Kallikrein 11 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Hippostasin, Anti-KLK11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~35 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... KLK11(11012)
mouse ... Klk11(56538)
rat ... Klk11(292849)

General description

The gene KLK11 (kallikrein 11) encodes a member of the kallikrein gene family. The gene is mapped to human chromosome 19q13.4. It is found to be highly expressed in the prostate. It has been found to be present in biological fluids and in serum of patients with various cancers. KLK11 is also found to be expressed in stomach, trachea, skin, and colon.

Immunogen

synthetic peptide corresponding to residues 68-82 of the human Kallikrein 11. This sequence is 86% identical in mouse and rat.

Biochem/physiol Actions

The gene KLK11 (kallikrein 11) encodes a putative serine protease that is overexpressed in prostate and ovarian cancer. It serves as a biomarker for prognosis of ovarian cancer. It can serve as a marker for distinguishing between prostate cancer and benign prostatic hyperplasia.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The usefulness of serum human kallikrein 11 for discriminating between prostate cancer and benign prostatic hyperplasia.
Nakamura T
Cancer Research, 63, 6543-6546 (2003)
Human kallikrein 11: a new biomarker of prostate and ovarian carcinoma.
Diamandis EP
Cancer Research, 62, 295-300 (2002)
Human kallikrein 11: an indicator of favorable prognosis in ovarian cancer patients.
Diamandis EP
Clinical Biochemistry, 37, 823-829 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service