Skip to Content
Merck
All Photos(1)

Key Documents

I9764

Sigma-Aldrich

Anti-Human IgG (γ-chain specific) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
R$2,157.00

R$2,157.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Select a Size

Change View
2 ML
R$2,157.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

R$2,157.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

immunoelectrophoresis: suitable
indirect ELISA: 1:50,000
quantitative precipitin assay: 2.0-3.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgG (immunoglobulin G) antibody subtype is a predominant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. IgG is found in four subclasses IgG1, IgG2, IgG3 and IgG4. IgG has a monomeric structure.

Application

Anti-Human IgG (γ-chain specific) antibody produced in rabbit has been used in western blotting.
Anti-Human IgG (γ-chain specific) antibody produced in rabbit was conjugated with BSA and coated over a nanofibrillar cellulose film to develop a platform for immunoassays and diagnostics.

Biochem/physiol Actions

IgG (immunoglobulin G) antibody participates in complement fixation and opsonization.
IgG antibody subtype provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein disulfide isomerase does not control recombinant IgG4 productivity in mammalian cell lines
Hayes NVL, et al.
Biotechnology and Bioengineering, 105(4), 770-779 (2010)
Comparison of antibody molecules produced from two cell lines with contrasting productivities and aggregate contents
Ishii Y, et al.
Biological & Pharmaceutical Bulletin, b14-00729 (2014)
James D Budge et al.
Data in brief, 29, 105217-105217 (2020-02-20)
The data presented in this article relates to the manuscript entitled 'Engineering of Chinese hamster ovary cell lipid metabolism results in an expanded ER and enhanced recombinant biotherapeutic protein production', published in the Journal Metabolic Engineering [1]. In the article
Tanya J Knight et al.
Biotechnology and bioengineering, 119(12), 3408-3420 (2022-09-10)
Mammalian cells, particularly Chinese hamster ovary cells, are the dominant system for the production of protein-based biotherapeutics, however, product degradation, particularly of Fc-fusion proteins, is sometimes observed that impacts the quality of the protein generated. Here, we identify the site
Hannes Orelma et al.
Biointerphases, 7(1-4), 61-61 (2012-10-12)
We introduce a new method to modify films of nanofibrillated cellulose (NFC) to produce non-porous, water-resistant substrates for diagnostics. First, water resistant NFC films were prepared from mechanically disintegrated NFC hydrogel, and then their surfaces were carboxylated via TEMPO-mediated oxidation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service