Skip to Content
Merck
All Photos(2)

Key Documents

I2909

Sigma-Aldrich

Indoramin hydrochloride

≥98% (HPLC), solid

Synonym(s):

N-[1-[2-(1H-Indol-3-yl)ethyl]-4-piperidinyl] benzamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$1,090.00
50 MG
R$4,529.00

R$1,090.00


Estimated to ship onApril 16, 2025



Select a Size

Change View
10 MG
R$1,090.00
50 MG
R$4,529.00

About This Item

Empirical Formula (Hill Notation):
C22H26ClN3O
CAS Number:
Molecular Weight:
383.91
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,090.00


Estimated to ship onApril 16, 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: ~24 mg/mL
H2O: insoluble

originator

Wyeth

SMILES string

[Cl].O=C(NC1CCN(CC1)CCc2c[nH]c3ccccc23)c4ccccc4

InChI

1S/C22H25N3O.Cl/c26-22(17-6-2-1-3-7-17)24-19-11-14-25(15-12-19)13-10-18-16-23-21-9-5-4-8-20(18)21;/h1-9,16,19,23H,10-15H2,(H,24,26);

InChI key

NZEVFUHXTSASST-UHFFFAOYSA-N

Biochem/physiol Actions

α1-adrenoceptor antagonist.

Features and Benefits

This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service