Skip to Content
Merck
All Photos(1)

Key Documents

I2032

Sigma-Aldrich

Monoclonal Anti-Insulin-Like Growth Factor Binding Protein-1 antibody produced in mouse

clone 33627.11, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IGFBP-1

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
R$7,296.00

R$7,296.00


Estimated to ship onMay 09, 2025



Select a Size

Change View
500 μG
R$7,296.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

R$7,296.00


Estimated to ship onMay 09, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

33627.11, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: suitable
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGFBP1(3484)

General description

The effects of insulin-like growth factors (IGF) are modulated by a family of insulin-like growth factor binding proteins 1-6. (IGFBPs -1-6). IGFBP-1 is secreted as an endocrine factor by liver, kidneys, decidua and in human amniotic fluid, but has the ability to signal in autocrine and paracrine fashion. Post-translational modifications such as ubiquitination, glycosylation and phosphorylation alter the activity of IGFBPs. Phosphorylation of IGFBP-1 is mainly mediated by Casein kinase I and II and cAMP-dependent protein kinases. IGFBP-1 alters the bioavailability of IGF in the serum by binding and sequestration so that a regulated amount id available to act on the target cells. The functions of IGFBP-1 are most important in the physiology of female reproductive system in mammals, for growth and development of the developing fetus
Monoclonal Anti-Human Insulin-like Growth Factor Binding Protein-1 (IGFBP-1) recognizes recombinant human IGFBP-1. No cross-reactivity is seen with recombinant human IGFBP-2, IGFBP-3, and IGFBP-4.

Specificity

The biological activity is measured by its ability to neutralize recombinant human IGFBP-1 in the presence of recombinant human IGF-I.

Immunogen

purified recombinant human insulin-like growth factor binding protein 1, expressed in E. coli.

Application

Anti- Insulin-Like Growth Factor Binding Protein-1 antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. For ELISA, a working concentration of 2 μg/ml may be used. The antibody is suitable for neutralization reactions (ND50 is 10-40 μg/ml).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Fowler et al.
Human reproduction update, 6(5), 495-504 (2000-10-25)
Insulin-like growth factor-1 (IGFBP-1) is particularly important in human female reproductive physiology, where it is involved with other factors in a complex system which regulates menstrual cycles, puberty, ovulation, decidualization, implantation and fetal growth. This has implications for clinical obstetrics
Terry J Smith
Pharmacological reviews, 62(2), 199-236 (2010-04-16)
This topically limited review explores the relationship between the immune system and insulin-like growth factors (IGF-I and IGF-II) and the proteins through which they act, including IGF-I receptor (IGF-IR) and the IGF-I binding proteins. The IGF/IGF-IR pathway plays important and
K M Kelley et al.
The international journal of biochemistry & cell biology, 28(6), 619-637 (1996-06-01)
The IGFBPs are a family of homologous proteins that have co-evolved with the IGFs and that confer upon the IGF regulatory system both functional and tissue specificity. IGFBPs are not merely carrier proteins for IGFs, but hold a central position
M Westwood
Reviews of reproduction, 4(3), 160-167 (1999-10-16)
Insulin-like growth factors and their binding proteins are key regulators of fetal and maternal tissue growth and development during human pregnancy. Insulin-like growth factors, particularly IGF-II, are produced in abundance by the trophoblast cells of the placenta, whereas one of
Ali Vaziri-Gohar et al.
Molecular and cellular endocrinology, 422, 160-171 (2015-12-23)
Tamoxifen, a selective estrogen receptor modulator, is a commonly prescribed adjuvant therapy for estrogen receptor-α (ERα)-positive breast cancer patients. To determine if extracellular factors contribute to the modulation of IGF-1 signaling after tamoxifen treatment, MCF-7 cells were treated with IGF-1 in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service