Skip to Content
Merck
All Photos(1)

Key Documents

I1388

Sigma-Aldrich

(−)-Indolactam V

~90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N3O2
CAS Number:
Molecular Weight:
301.38
MDL number:
UNSPSC Code:
12352103

Assay

~90%

storage temp.

−20°C

SMILES string

CC(C)[C@@H]1N(C)c2cccc3[nH]cc(C[C@@H](CO)NC1=O)c23

Application

Synthetic activator of protein kinase C.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endo, Y., et al.
Tetrahedron Letters, 42, 5905-5905 (1986)
Kyle P Glover et al.
PloS one, 10(10), e0139850-e0139850 (2015-10-03)
Activation of stress response pathways in the tumor microenvironment can promote the development of cancer. However, little is known about the synergistic tumor promoting effects of stress response pathways simultaneously induced in the tumor microenvironment. Therefore, the purpose of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service