Skip to Content
Merck
All Photos(1)

Key Documents

I123

Sigma-Aldrich

ICI 216,140

>95%, solid

Synonym(s):

(CH3)2CHCO-His-Trp-Ala-Val-D-Ala-His-Leu-NHCH3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H65N13O8
CAS Number:
Molecular Weight:
916.08
MDL number:
UNSPSC Code:
12352200

Assay

>95%

form

solid

color

white

storage temp.

−20°C

SMILES string

CNC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@@H](C)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4c[nH]cn4)NC(=O)C(C)C)C(C)C

Biochem/physiol Actions

Potent in vivo receptor antagonist of the bombesin/gastrin releasing peptide receptor.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B J Campbell et al.
Regulatory peptides, 49(3), 249-255 (1994-01-13)
The mechanisms of action of gastrin and gastrin releasing peptide (GRP) in stimulating gastric acid secretion were examined in the anaesthetized turkey. Gastrin and GRP produced dose-dependent increases in acid secretion that were inhibited by the gastrin/CCK-B antagonist CI988. The
ICI 216140 and other potent in vivo antagonist analogs of bombesin/gastrin-releasing peptide.
Camble, R., et al.
Peptides: Chemistry, Structure and Biology, Proceedings of the Eleventh American Peptide Symposium, 174-174 (1989)
Branimir Zogovic et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 301(5), R1486-R1494 (2011-08-19)
Bombesin, a 14 amino-acid peptide, is pressor when administered intravenously in rat and pressor and sympathoexcitatory when applied intracerebroventricularly. To determine the spinal effects of bombesin, the peptide was administered acutely in the intrathecal space at around thoracic spinal cord
B Campbell et al.
The American journal of physiology, 261(1 Pt 1), G16-G21 (1991-07-01)
Endocrine cells in the acid-secreting part of the avian stomach, the proventriculus, contain two forms of gastrin-releasing peptide (GRP) of 27 and 6 residues, respectively. We have examined the actions of exogenous GRP-27 and GRP-6 and endogenously released GRP in
R Camble et al.
Life sciences, 45(17), 1521-1527 (1989-01-01)
The GRP receptor mediated growth response in Swiss 3T3 cells has been used to identify BN/GRP antagonists. Analysis of bombesin antagonism by substance P analogues and by truncated GRP analogues revealed that deletion of the C-terminal methionine residue was important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service