Skip to Content
Merck
All Photos(1)

Key Documents

HS2201A

Droplet phlebotomy tray inserts for 13 mm test tube rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.35
Pricing and availability is not currently available.

feature

Holds 13 mm Tube Rack,

packaging

pack of 4 inserts

manufacturer/tradename

Heathrow Scientific HS2201A

General description

Utility Tray Insert A for 13mm test tube rack with eight compartments.

Legal Information

Droplet is a trademark of Heathrow Scientific LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service