Key Documents
HS120656
Dual Purpose Scoops
size large, white, non-sterile
Select a Size
100 EACH
R$167.00
R$167.00
Estimated to ship onMay 24, 2025
Select a Size
Change View
100 EACH
R$167.00
About This Item
UNSPSC Code:
41122403
NACRES:
NB.71
R$167.00
Estimated to ship onMay 24, 2025
material
polystyrene
white
size
large
sterility
non-sterile
packaging
pack of 100 ea
manufacturer/tradename
Heathrow Scientific 120656
L × W × H
127 mm × 88.9 mm × 0.5 mm
Related Categories
General description
The dual purpose scoop is a unique economical solution that combines the function of a scoop and a weigh boat into oneproduct. The weighing process has been streamlined, now just sample or retrieve the material in the scoop and place it directly on the scale. This eliminates the need for a weigh boat or weighing paper and saves valuable space in the lab. When finished the dual solution scoop is disposable.
Features and Benefits
  • Made from anti-static polystyrene that is waterproof andreduces the chance of materials sticking to the scoop
  • Ships and stores flat and when ready to use, is easy to assemble by inserting the locking tab into the hole
  • Locking tab feature enables a scoopshape to help retain materials
  • Coated to stand up to powders, gels, and semi-solids
  • Broad tip design keeps the scoop stable while on the scale and enables materialsto be easily scooped and dispensed
  • Disposable
Choose from one of the most recent versions:
Certificates of Analysis (COA)