Key Documents
HS120098
Pearl Pipette
adjustable, volume 0.5-10 μL, light blue, single-channel
Select a Size
1 EACH
R$1,181.00
R$1,181.00
Check Cart for Availability
Select a Size
Change View
1 EACH
R$1,181.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
R$1,181.00
Check Cart for Availability
material
light blue
white
feature
adjustable
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120098
volume measuring range
0.5-10 μL
General description
The Pearl is a single-channel pipette offering adjustable volume and precision liquid handling. The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.
Features and Benefits
  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digit volume display
  • Color coded and labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified and Manufactured in accordance with ISO08655 and GLP regulations
  • Autoclavable
Legal Information
Pearl is a trademark of Specac, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Dear Sir/Madam We purchased a Pearl Pipette (HS120105 adjustable, volume 500-5000 μL, pink, single-channel), Could you please help me find out the proper pipette tip pack for the same?
    1 answer
    1. This is a product of Heathrow Scientific. Unfortunately, the manufacturer does not offer pipette tips and makes no recommendation as to compatibility with other brands.
      Helpful?