Skip to Content
Merck
All Photos(3)

Documents

HS10050

Gusto® High Speed Mini Centrifuge

AC/DC input 100 V / 240 V, universal plug set, blue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701
NACRES:
NB.11

material

blue

feature

to hold 12 x 1.5/2.0 mL tubes
to hold 16 x 0.2 mL tubes (single or 2x8 strip)
to hold 32 x 0.2 mL tubes (singles, or 4 x 8 Strip)
to hold 6 x 1.5/2 mL tubes

packaging

pkg of 1 ea

manufacturer/tradename

Heathrow Scientific HS10050

parameter

12,500 rpm speed (max.)
2000 × g max. RCF (max.)
6,000 rpm max. RCF
9,783 × g max. RCF

General description

Maximum Speed with Minimal Space
This compact centrifuge is ideal for protocols calling for fast spins with a higher RCF requirement.
Ideal for PCR, microfilter cell separation, clinical & diagnostic applications, and HPLC protocols.

Features and Benefits

  • Made of durable ABS with UV-Resistant Coating
  • Brush-less motor for quiet operation and low vibration
  • Quick-spin button for momentary spin requirements, press and hold to operate, release to end spin cycle
  • Tool Free rotor exchange
  • Safety features: Sample imbalance, tilt auto-shut off and interlock lid prevents opening while rotor is turning

Packaging

INCLUDES:
Gusto® High-Speed Mini-Centrifuge
12-Place 1.5/2.0 mL Tube Rotor
32-Place 0.2 mL Tube Rotor (singles, or 4 x 8 Strip)
12 Tube Adapters 0.2 mL
12 Tube Adapters 0.5 mL
Low Voltage, Double Insulated, power adapter 110–240V 50/60 Hz, with 4 interchangeable cord types

Legal Information

GUSTO is a registered trademark of Heathrow Scientific

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service