Skip to Content
Merck
All Photos(2)

Key Documents

HPA028543

Sigma-Aldrich

Anti-OTUD3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DUBA4, Anti-KIAA0459, Anti-OTU domain containing 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.4 μg/mL
immunofluorescence: 1-4 μg/mL

immunogen sequence

KGMDSEDDLRDEVEDAVQKVCNATGCSDFNLIVQNLEAENYNIESAIIAVLRMNQGKRNNAEENLEPSGRVLKQCGP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OTUD3(23252)

Immunogen

Recombinant protein corresponding to OTU deubiquitinase 3.

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77062

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peiyi Xie et al.
Aging, 13(15), 19317-19338 (2021-08-13)
OTU domain-containing protein 3 (OTUD3), a deubiquitinating enzyme, has been shown to participate in progression of multiple malignancies. The accurate function of OTUD3 in hepatocellular carcinoma (HCC) progression remains elusive. We found that OTUD3 was significantly overexpressed in HCC, and
Pengfei Zhang et al.
Cell death and differentiation, 27(11), 3177-3195 (2020-06-03)
Ovarian tumour domain-containing protein 3 (OTUD3), a key OTU (ovarian tumour protease) family deubiquitylase, plays context-dependent roles in cancers. It suppresses tumorigenesis in breast, colon, liver and cervical cancer through stabilizing PTEN (phosphatase and tension homologue deleted on chromosome 10)
Yonghui Zhang et al.
Cell death & disease, 14(6), 378-378 (2023-06-28)
The ubiquitin-proteasome system (UPS) controls protein turnover, and its dysfunction contributes to human diseases including cancer. Deubiquitinating enzymes (DUBs) remove ubiquitin from proteins to maintain their stability. Inhibition of DUBs could induce the degradation of selected oncoproteins and has therefore
Tongde Du et al.
Nature communications, 10(1), 2914-2914 (2019-07-04)
The deubiquitylase OTUD3 plays a suppressive role in breast tumorigenesis through stabilizing PTEN protein, but its role in lung cancer remains unclear. Here, we demonstrate that in vivo deletion of OTUD3 indeed promotes breast cancer development in mice, but by contrast
Fengju Jia et al.
Cell death & disease, 13(4), 418-418 (2022-05-01)
Iron deposits are neuropathological hallmark of Parkinson's disease (PD). Iron regulatory protein 2 (IRP2) is a key factor in regulating brain iron homeostasis. Although two ubiquitin ligases that promote IRP2 degradation have been identified, the deubiquitylase for stabilization of IRP2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service