Skip to Content
Merck
All Photos(5)

Key Documents

HPA025729

Sigma-Aldrich

Anti-NRARP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Notch-regulated ankyrin repeat-containing protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MSQAELSTCSAPQTQRIFQEAVRKGNTQELQSLLQNMTNCEFNVNSFGPEGQTALHQSVIDGNLELVKLLVKFGADIRLANRDGWSALHIAAF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NRARP(441478)

General description

NRARP (NOTCH-regulated ankyrin repeat protein) works as a negative feedback regulator in Notch (neurogenic locus notch homolog protein) signaling. On the other hand, expression of NRARP is controlled by Notch protein. The protein has two ankyrin repeats.

Immunogen

Notch-regulated ankyrin repeat-containing protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

During development, NRARP (NOTCH-regulated ankyrin repeat protein) is involved in crosstalk between NOTCH (neurogenic locus notch homolog protein) and WNT (wingless-type mouse mammary tumor virus integration site) signaling. Presence of NRARP allows proper vessel density in angiogenesis. In breast cancer, NRARP might enhances the cell proliferation. It is upregulated in thyroid cancer tissues and is associated with cell growth and invasion. It is also overexpressed in hepatocellular carcinoma tumor tissues.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75430

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufeng Liao et al.
Oncology letters, 16(2), 1885-1891 (2018-07-17)
Notch-regulated ankyrin-repeat protein (NRARP) has recently been reported to be involved in a number of malignant cancers; however, its role in non-small lung cancer (NSCLC) remains unclear. The present study aimed to identify whether NRARP could be applied as a
Overexpression of NOTCH-regulated ankyrin repeat protein is associated with breast cancer cell proliferation.
Imaoka T, et al.
Anticancer Research, 34, 2165-2171 (2014)
Nrarp coordinates endothelial Notch and Wnt signaling to control vessel density in angiogenesis.
Phng LK, et al.
Developmental Cell, 16, 70-82 (2009)
Downregulation of Notch-regulated Ankyrin Repeat Protein Exerts Antitumor Activities against Growth of Thyroid Cancer.
Chu BF, et al.
Chinese Medical Journal (English Edition), 129, 1544-1552 (2016)
Pingping Zhu et al.
Nature communications, 6, 7122-7122 (2015-05-20)
Liver cancer stem cells (CSCs) harbour self-renewal and differentiation properties, accounting for chemotherapy resistance and recurrence. However, the molecular mechanisms to sustain liver CSCs remain largely unknown. In this study, based on analysis of several hepatocellular carcinoma (HCC) transcriptome datasets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service