Skip to Content
Merck
All Photos(6)

Key Documents

HPA025287

Sigma-Aldrich

Anti-FBLIM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FBLP-1, Anti-Filamin-binding LIM protein 1, Anti-MIG2-interacting protein, Anti-Migfilin, Anti-Mitogen-inducible 2-interacting protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

CEPCYQDTLERCGKCGEVVRDHIIRALGQAFHPSCFTCVTCARCIGDESFALGSQNEVYCLDDFYRKFAPVCSICENPIIPRDGKDAFKIECMGRNFHENCYRCEDCRILLSVEPTDQGCYPLNNHLFCKPCHV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBLIM1(54751)

General description

FBLIM1 (filamin binding LIM protein 1) is present at the cell-cell adhesion areas. The protein has the amino-terminal, proline rich-region and carboxy-terminal LIM (LIN-11, ISL-1 and MEC-3) domains.[1] It is commonly referred to as migfilin.

Immunogen

Filamin-binding LIM protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FBLIM1 (filamin binding LIM protein 1) plays an important role in association between the cell membrane and the actin cytoskeleton. It mainly interacts with MIG-2 (mitogen-inducible gene 2 protein), filamin and VASP (vasodilator-stimulated phosphoprotein), thus controlling cell shape and movements. In cardiomyocytes, it interacts with transcription factor CSX/NKX2-5 (cardiac-specific homeobox) and enhances the differentiation. The FBLIM1 gene is upregulated in osteoarthritis chondrocytes and might be involved with osteoarthritis pathogenesis. In various carcinoma cells, the FBLIM1-associated signaling in disrupted which leads to abnormal Src activation and anoikis resistance in cells. This gene is downregulated in breast cancer cells. In esophageal cancer cells, it suppresses invasion partly by enhancing degradation of β-catenin. However, in glioma cells FBLIM1 promotes migration as well as invasion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70103

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanako Ishizuka et al.
Journal of neuroscience research, 96(5), 789-802 (2017-11-09)
Migfilin, encoded by FBLIM1 at the 1p36 locus, is a multi-domain adaptor protein essential for various cellular processes such as cell morphology and migration. Small deletions and duplications at the 1p36 locus, monosomy of which results in neurodevelopmental disorders and
Mitali Das et al.
PloS one, 6(10), e26355-e26355 (2011-11-02)
Cell adhesion and migration depend on engagement of extracellular matrix ligands by integrins. Integrin activation is dynamically regulated by interactions of various cytoplasmic proteins, such as filamin and integrin activators, talin and kindlin, with the cytoplasmic tail of the integrin
Migfilin interacts with Src and contributes to cell-matrix adhesion-mediated survival signaling.
Zhao J, et al.
The Journal of Biological Chemistry, 284, 34308-34320 (2009)
Migfilin's elimination from osteoarthritic chondrocytes further promotes the osteoarthritic phenotype via ?-catenin upregulation.
Gkretsi V, et al.
Biochemical and Biophysical Research Communications, 430, 494-499 (2013)
Migfilin regulates esophageal cancer cell motility through promoting GSK-3?-mediated degradation of ?-catenin.
He H, et al.
Molecular Cancer Research, 10, 273-281 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service