Skip to Content
Merck
All Photos(3)

Key Documents

HPA024829

Sigma-Aldrich

Anti-PLEKHF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DNB5, Anti-FLJ13187, Anti-PHAFIN2, Anti-ZFYVE18, Anti-pleckstrin homology domain containing, family F (with FYVE domain) member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

VYGNIVIQKKKYNKQHIIPLENVTIDSIKDEGDLRNGWLIKTPTKSFAVYAATATEKSEWMNHINKCVTDLLSKSGKTPSN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLEKHF2(79666)

General description

The gene PLEKHF2 (pleckstrin homology and FYVE domain containing 2) is mapped to human chromosome 8q22. The encoded protein belongs to the Phafin protein family and contains pleckstrin homology (PH) domain and a FYVE (Fab 1, YOTB, Vac 1, and EEA1) domain. It is present in the early endosomes and endoplasmic reticulum.

Immunogen

pleckstrin homology domain containing, family F (with FYVE domain) member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PLEKHF2 (pleckstrin homology and FYVE domain containing 2) regulates the structure and function of endosomes with the help of Ras-related protein Rab5. It is also involved in EGFR (epidermal growth factor receptor) degradation. PLEKHF2 interacts with PI3P (phosphatidylinositol-3-phosphate) and controls the induction of autophagy. It is expressed in human hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76388

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kia Wee Tan et al.
Journal of cell science, 134(14) (2021-06-11)
Macropinocytosis allows cells to take up extracellular material in a non-selective manner into large vesicles called macropinosomes. After internalization, macropinosomes acquire phosphatidylinositol 3-phosphate (PtdIns3P) on their limiting membrane as they mature into endosomal-like vesicles. The molecular mechanisms that underlie recycling
Changfei Li et al.
Journal of molecular medicine (Berlin, Germany), 86(4), 471-484 (2008-02-22)
We recently identified the Phafin protein family, whose members all contain an N-terminal PH domain (pleckstrin homology) and a C-terminal FYVE (Fab1, YGLO23, Vps27, and EEA1) domain. LAPF (lysosome-associated apoptosis-inducing protein containing PH and FYVE domains, also known as Phafin-1)
Kay Oliver Schink et al.
Nature communications, 12(1), 6577-6577 (2021-11-14)
Uptake of large volumes of extracellular fluid by actin-dependent macropinocytosis has an important role in infection, immunity and cancer development. A key question is how actin assembly and disassembly are coordinated around macropinosomes to allow them to form and subsequently
Nina Marie Pedersen et al.
Traffic (Copenhagen, Denmark), 13(11), 1547-1563 (2012-07-24)
Phosphatidylinositol 3-phosphate (PtdIns3P) orchestrates endosomal cargo transport, fusion and motility by recruiting FYVE or PX domain-containing effector proteins to endosomal membranes. In an attempt to discover novel PtdIns3P effectors involved in the termination of growth factor receptor signalling, we performed
Jie Hu et al.
PloS one, 6(10), e26003-e26003 (2011-10-15)
Recurrence prediction of hepatitis B virus (HBV)-related hepatocellular carcinoma (HCC) patients undergoing liver transplantation (LT) present a great challenge because of a lack of biomarkers. Genetic variations play an important role in tumor development and metastasis. Oligonucleotide microarrays were used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service