Skip to Content
Merck
All Photos(2)

Key Documents

HPA024828

Sigma-Aldrich

Anti-XKR6 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-XK-related protein 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

YGVLHPTGPRAKILASSCCAELLWGIPLPPDVEPMAPEIPGYRGTQVTPTRAVTEQQEDLTADTCLPVFQVRPMGPPTPLGRPYLPEGPLIKIDMPR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XKR6(286046)

General description

The gene XKR6 (XK related 6) is mapped to human chromosome 8p23.1. The encoded protein belongs to the XKR family of proteins and is a transmembrane protein.

Immunogen

XK-related protein 6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In red blood cells, XKR6 (XK related 6) belongs to the Kell blood group of antigens. It might be involved in SLE (systemic lupus erythematosus).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76375

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Suzuki et al.
The Journal of biological chemistry, 289(44), 30257-30267 (2014-09-19)
Apoptotic cells expose phosphatidylserine (PtdSer) on their surface as an "eat me" signal. Mammalian Xk-related (Xkr) protein 8, which is predicted to contain six transmembrane regions, and its Caenorhabditis elegans homolog CED-8 promote apoptotic PtdSer exposure. The mouse and human
Elisa Alonso-Perez et al.
PloS one, 7(9), e45356-e45356 (2012-10-11)
Systemic Lupus Erythematosus (SLE) shows a spectrum of clinical manifestations that complicate its diagnosis, treatment and research. This variability is likely related with environmental exposures and genetic factors among which known SLE susceptibility loci are prime candidates. The first published
M L Budarf et al.
Genes and immunity, 12(1), 51-58 (2010-10-22)
Systemic lupus erythematosus (SLE) is a multisystem autoimmune disease. Multiple genetic and environmental factors contribute to the pathogenesis of this disease. Recent genome-wide association studies have added substantially to the number of genes associated with SLE. To replicate some of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service