Skip to Content
Merck
All Photos(4)

Documents

HPA024171

Sigma-Aldrich

Anti-PPP1R12B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Myosin phosphatase target subunit 2, Anti-Myosin phosphatase-targeting subunit 2, Anti-Protein phosphatase 1 regulatory subunit 12B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GTGINFWTKDEDETDGSEEVKETWHERLSRLESGGSNPTTSDSYGDRASARARREAREARLATLTSRVEEDSNRDYKKLYESALTENQKLKTKLQEAQLELADIKS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP1R12B(4660)

General description

The gene PPP1R12B (protein phosphatase 1 regulatory subunit 12B) is mapped to human chromosome 1q32.1. PPP1R12B is expressed in the heart, skeletal muscle and brain.

Immunogen

Protein phosphatase 1 regulatory subunit 12B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PPP1R12B (protein phosphatase 1 regulatory subunit 12B) is associated with childhood asthma. It is a part of the myosin phosphatase complex. The complex controls contractile responses in muscle and nonmuscle cells. PPP1R12B is upregulated in disorders, including ischaemia, atrial fibrillation, myocardial infarction and cardiomyopathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76534

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of PPP1R12B as a candidate gene for childhood asthma in Russians.
Maxim B Freidin et al.
Journal of genetics, 92(1), 93-96 (2013-05-04)
M Fujioka et al.
Genomics, 49(1), 59-68 (1998-05-08)
Myosin phosphatase target subunit 1 (MYPT1), a subunit of myosin phosphatase, plays a pivotal role in the regulation of myosin phosphatase activity. Here we have cloned a novel isoform of MYPT1, termed MYPT2, from a human brain cDNA library screened

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service