Skip to Content
Merck
All Photos(7)

Key Documents

HPA023605

Sigma-Aldrich

Anti-UBE2O antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-E2-230K, Anti-ubiquitin-conjugating enzyme E2O

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Estimated to ship onApril 06, 2025



Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Estimated to ship onApril 06, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GVKQHVKETKLKLEDRSVVPRDVVRHMRSTDSQCGTVIDVNIDCAVKLIGTNCIIYPVNSKDLQHIWPFMYGDYIAYDCWLGKVYDLKNQIILKLSNGARCSMNTEDGAKLYDVCPHVSDSGLFF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2O(63893)

General description

The gene UBE2O (ubiquitin conjugating enzyme E2 O) encodes a member of the E2 family of ubiquitin-conjugating enzymes. The encoded protein is large with a molar mass of 140kDa. It contains a ubiquitin-conjugating (UBC) domain and is expressed ubiquitously, with increased expression in brain, skeletal muscle, and heart tissues. It is found to be up-regulated during the reticulocyte stage of erythroid differentiation.

Immunogen

ubiquitin-conjugating enzyme E2O recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The E2 enzyme encoded by the gene UBE2O (ubiquitin conjugating enzyme E2 O) negatively regulates TRAF6-mediated IL-1R (interleukin-1R)/TLR4 (toll-like receptor4) signaling via the inhibition of polyubiquitination of TRAF6 (tumor necrosis factors receptor associated factor 6). It hinders the formation of MyD88-TRAF6 protein complex. It prevents TRAF6-dependent NF-κB (nuclear factor-κB) signaling. It is involved in the ubiquitination of the nuclear localization signal of BAP1 (BRCA1 associated protein 1), a tumor suppressor that regulates cell proliferation. UBE2O interacts and monoubiquitinates SMAD6, an inhibitory regulator of bone morphogenetic protein (BMP)/SMAD signaling, thereby regulating BMP7 (bone morphogenetic protein)-induced adipogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86463

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autodeubiquitination protects the tumor suppressor BAP1 from cytoplasmic sequestration mediated by the atypical ubiquitin ligase UBE2O
Mashtalir N
Molecular Cell, 54, 392-406 (2014)
Xiaofei Zhang et al.
The EMBO journal, 32(7), 996-1007 (2013-03-05)
SMAD6 is a crucial feedback inhibitory regulator of bone morphogenetic protein (BMP)/SMAD signalling. Although little is known regarding the post-transcriptional modification of inhibitory SMADs and the mechanism by which their function is regulated. In this study, using a whole proteomic
Xiaofei Zhang et al.
Cell research, 23(3), 366-377 (2013-02-06)
Tumor necrosis factor (TNF) receptor-associated factor 6 (TRAF6) is a key regulator of the activation of transcription factor NF-κB by the interleukin-1 receptor (IL-1R)/Toll-like receptor (TLR) superfamily. Recruitment of TRAF6 to the receptor-associated IRAK1-IRAK4-MyD88 adaptor protein complex induces lysine 63

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service