Skip to Content
Merck
All Photos(3)

Key Documents

HPA023567

Sigma-Aldrich

Anti-MSL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-MSL-1, Anti-MSL1-like 1, Anti-Male-specific lethal 1 homolog, Anti-Male-specific lethal 1-like 1, Anti-Male-specific lethal-1 homolog 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Estimated to ship onMay 02, 2025



Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Estimated to ship onMay 02, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

FGSTERKTPVKKLAPEFSKVKTKTPKHSPIKEEPCGSLSETVCKRELRSQETPEKPRSSVDTPPRLSTPQKGPSTHPKEKAFSSEIEDLPYLSTTEMY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MSL1(339287)

General description

The gene MSL1 (male specific lethal 1 homolog) encodes a histone acetyl transferase-associated protein that spans a length of 614 amino acid residues. It does not contain any globular domains.

Immunogen

Male-specific lethal 1 homolog recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene MSL1 (male specific lethal 1 homolog) encodes a protein associated with mammalian MOF (male-absent-on-the-first)-MSL (male-specific-lethal) complex that is also referred to as the ‘dosage compensation complex′ (DCC). This complex is involved in the equalization of X-chromosomal genes expression between males and females. It is found to form a complex with Nupr1 (nuclear protein 1) in the nucleus in response to DNA-damage and may be involved in DNA repair activity. MSL1 protein is found to regulate acetylation of global histone H4 via its interaction with KAT8 (lysine acetyltransferase 8).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75963

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Aguado-Llera et al.
PloS one, 8(10), e78101-e78101 (2013-11-10)
The stress protein Nupr1 is a highly basic, multifunctional, intrinsically disordered protein (IDP). MSL1 is a histone acetyl transferase-associated protein, known to intervene in the dosage compensation complex (DCC). In this work, we show that both Nupr1 and MSL1 proteins
Ruslan I Dmitriev et al.
Journal of cellular biochemistry, 115(11), 1967-1973 (2014-06-11)
MSL1 protein regulates global histone H4 acetylation at residue K16 in stem and cancer cells, through interaction with KAT8. The functional significance of mammalian MSL1 isoforms, involved in various protein interactions, is poorly understood. We report the identification of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service