Skip to Content
Merck
All Photos(6)

Key Documents

HPA022034

Sigma-Aldrich

Anti-MPRIP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-M-RIP, Anti-Myosin phosphatase Rho-interacting protein, Anti-RIP3, Anti-Rho-interacting protein 3, Anti-p116Rip

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

R$4,567.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RVEGHVGELGDFQVKNSQALMCLENCREQLRSLPRASQEDEQDARAASLASVESALVSAIQALQHWPAPAHGGARAQLETGGTEENGKPASLQQCSQSELTEQEQVRLLSDQIALEASLISQIADSLKNTT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPRIP(23164)

General description

MPRIP (Myosin phosphatase Rho interacting protein) is a serine/threonine protein kinase mapped on human chromosome 17p11.2. It is localized to actin-myosin stress fibers.[1]

Immunogen

Myosin phosphatase Rho-interacting protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MPRIP (Myosin phosphatase Rho interacting protein) controls the switch from apoptosis to necroptosis. It is associated with several diseases. MPRIP functions as a component of the myosin phosphatase complex and combines with the myosin binding subunit of myosin phosphatase and RhoA. Absence or deficiency of MPRIP protects hepatocytes from ethanol-mediated injury, prevents cerulean-activated acute necrotizing pancreatitis, suppresses photoreceptor and cone cell death, and improves macrophage necrosis in advanced atherosclerosis lesions.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75694

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Mosca-Boidron et al.
Clinical genetics, 82(1), 41-47 (2011-07-05)
Most microdeletion syndromes identified before the implementation of array-comparative genomic hybridization (array-CGH) were presumed to be well-defined clinical entities. However, the introduction of whole-genome screening led not only to the description of new syndromes but also to the recognition of
Howard K Surks et al.
The Journal of biological chemistry, 280(52), 42543-42551 (2005-11-01)
Vascular smooth muscle cell contraction and relaxation are directly related to the phosphorylation state of the regulatory myosin light chain. Myosin light chains are dephosphorylated by myosin phosphatase, leading to vascular smooth muscle relaxation. Myosin phosphatase is localized not only
J-X Li et al.
Cell death & disease, 5, e1278-e1278 (2014-06-06)
Receptor-interacting protein (RIP)3 is a critical regulator of necroptosis and has been demonstrated to be associated with various diseases, suggesting that its inhibitors are promising in the clinic. However, there have been few RIP3 inhibitors reported as yet. B-Raf(V600E) inhibitors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service