Skip to Content
Merck
All Photos(2)

Key Documents

HPA020255

Sigma-Aldrich

Anti-MPDZ antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Multi-PDZ domain protein 1, Anti-Multiple PDZ domain protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

R$4,567.00


Estimated to ship onJune 02, 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

VVNILKELPIEVTMVCCRRTVPPTTQSELDSLDLCDIELTEKPHVDLGEFIGSSETEDPVLAMTDAGQSTEEVQAPLAMWEAGIQHIELEKGSKGLGFS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPDZ(8777)

General description

Multiple PDZ domain protein (MPDZ) is a 220kDa protein expressed in the borders of epithelial cells and tight junctions. It possesses 13 PDZ (PSD95, Dlg1 and zo-1) binding domains.

Immunogen

Multiple PDZ domain protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Multiple PDZ domain protein (MPDZ) interacts with various subunits of dopamine, serotonin and γ-amino butyric acid receptors. It has a role in the N-methyl-D-aspartate (NMDA)-mediated α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) trafficking cascade. MPDZ also binds to many signaling molecules and scaffolding proteins.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75199

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuan Ouyang et al.
Journal of the peripheral nervous system : JPNS, 24(2), 195-206 (2019-05-24)
The blood-nerve barrier (BNB) formed by tight junction-forming endoneurial microvessels located in the innermost compartment of peripheral nerves, and the perineurium serve to maintain the internal microenvironment required for normal signal transduction. The specific molecular components that define the normal
Aleksandra Sindic et al.
American journal of physiology. Renal physiology, 297(1), F36-F45 (2009-05-08)
We previously found that the Ca(2+)-sensing receptor (CaR) interacts with and inactivates the inwardly rectifying K(+) channel Kir4.2 that is expressed in the kidney cortex and that has a COOH-terminal PDZ domain. To identify potential scaffolding proteins that could organize
Victor M Karpyak et al.
Addiction biology, 17(4), 798-806 (2011-07-19)
Model studies in mice indicate that the severity of alcohol withdrawal is associated with polymorphic variation and expression of the MPDZ gene. Current knowledge about variation in the human MPDZ gene is limited; however, our data indicate its potential association
Victor M Karpyak et al.
Alcoholism, clinical and experimental research, 33(4), 712-721 (2009-01-30)
Mpdz gene variations are known contributors of acute alcohol withdrawal severity and seizures in mice. To investigate the relevance of these findings for human alcoholism, we resequenced 46 exons, exon-intron boundaries, and 2 kilobases in the 5' region of the
Fabian Tetzlaff et al.
eLife, 7 (2018-04-06)
Angiogenesis is coordinated by VEGF and Notch signaling. DLL4-induced Notch signaling inhibits tip cell formation and vessel branching. To ensure proper Notch signaling, receptors and ligands are clustered at adherens junctions. However, little is known about factors that control Notch

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service