Skip to Content
Merck
All Photos(7)

Key Documents

HPA019777

Sigma-Aldrich

Anti-DAZL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-DAZ homolog, Anti-DAZ-like autosomal, Anti-Deleted in azoospermia-like, Anti-Deleted in azoospermia-like 1, Anti-SPGY-like-autosomal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

HEATPPSGNGPQKKSVDRSIQTVVSCLFNPENRLRNSVVTQDDYFKDKRVHHFRRSRAMLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DAZL(1618)

General description

DAZL (Deleted in azoospermia-like) is a SPGYLA encoded RNA binding protein belonging to the DAZ (deleted in azoospermia) gene family. In humans, it is expressed in germ cells and male gonad.

Immunogen

Deleted in azoospermia-like recombinant protein epitope signature tag (PrEST)

Application

Anti-DAZL antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

DAZL (Deleted in azoospermia-like) plays an important role in gametogenesis of both males and females. It is crucially important for spermatogenesis. It regulates translation by binding to the 3′-UTR of mRNA, specifically recognizing GUU triplets. Mutations in this gene may cause male infertility.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74469

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Bonache et al.
Andrology, 2(4), 596-606 (2014-05-08)
The molecular basis of spermatogenic failure (SpF) is still largely unknown. Accumulating evidence suggests that a series of specific events such as meiosis, are determined at the early stage of spermatogenesis. This study aims to assess the expression profile of
N N Chai et al.
Molecular human reproduction, 3(8), 705-708 (1997-08-01)
The DAZLA (DAZ Like Autosomal) gene on human chromosome 3 shares a high degree of homology with the DAZ (Deleted in AZoospermia) gene family on the Y chromosome, a gene family frequently deleted in males with azoospermia or severe oligospermia.
Z Shan et al.
Human molecular genetics, 5(12), 2005-2011 (1996-12-01)
We have isolated a series of human testis poly(A) cDNA clones by cross-hybridization to SPGY1, a Y gene homologous to DAZ. Their sequence analysis revealed an identical nucleotide composition in different 'full-length' clones, suggesting that all were encoded by the
R Saxena et al.
Nature genetics, 14(3), 292-299 (1996-11-01)
It is widely believed that most or all Y-chromosomal genes were once shared with the X chromosome. The DAZ gene is a candidate for the human Y-chromosomal Azoospermia Factor (AZF). We report multiple copies of DAZ (> 99% identical in
S Tsui et al.
Genomics, 65(3), 266-273 (2000-06-17)
The human DAZ (deleted in azoospermia) gene family on the Y chromosome and an autosomal DAZ-like gene, DAZL1, encode RNA-binding proteins that are expressed exclusively in germ cells. Their role in spermatogenesis is supported by their homology with a Drosophila

Articles

Stem cell markers, including embryonic stem cell, pluripotency, transcription factors, induced PSCs, germ cells, ectoderm, and endoderm markers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service