Skip to Content
Merck
All Photos(4)

Key Documents

HPA017732

Sigma-Aldrich

Anti-CNNM4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Ancient conserved domain- containing protein 4, Anti-Cyclin-M4, Anti-Metal transporter CNNM4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Estimated to ship onMay 29, 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

FSYYGTMALTSVPSDRSPAHPTPLSRSASLSYPDRTDVSTAATLAGSSNQFGSSVLGQYISDFSVRALVDLQYIKITRQQYQNGLLASRMENSPQFPIDGCTTHMENLAEKSELPVVDETTTLLNERNSL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CNNM4(26504)

General description

Ancient conserved domain-containing protein 4 (CNNM4) contains a highly conserved domain which is evolutionarily conserved from bacteria to mammals. In total, it has four transmembrane domains. CNNM4 is expressed in the gastrointestinal tract, neural retina and ameloblasts of the developing tooth. The gene encoding it is present on chromosome 2q11.

Immunogen

cyclin and CBS domain divalent metal cation transport mediator 4

Application

Anti-CNNM4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Ancient conserved domain-containing protein 4 (CNNM4) is involved in metal ion transport. It may remove unwanted Mg2+ ions which interfere with crystal formation in enamel and thus maintain the serum Mg2+ levels. In the epithelium, CNNM4 expels intracellular Mg2+ outside the cells in exchange for Na+. It binds to the enzyme phosphatase of regenerating liver (PRL) which is up-regulated in malignant human cancers. Along with PRL, CNNM4 is involved in energy metabolism and 5′ AMP-activated protein kinase (AMPK)/ mechanistic target of rapamycin (mTOR) signaling. Mutations in CNNM4 have been shown to be associated with recessive cone-rod dystrophy and amelogenesis imperfecta (Jalili syndrome).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73140

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Hirata et al.
The Journal of biological chemistry, 289(21), 14731-14739 (2014-04-08)
Ancient conserved domain protein/cyclin M (CNNM) family proteins are evolutionarily conserved Mg(2+) transporters. However, their biochemical mechanism of action remains unknown. Here, we show the functional importance of the commonly conserved cystathionine-β-synthase (CBS) domains and reveal their unique binding ability
David A Parry et al.
American journal of human genetics, 84(2), 266-273 (2009-02-10)
The combination of recessively inherited cone-rod dystrophy (CRD) and amelogenesis imperfecta (AI) was first reported by Jalili and Smith in 1988 in a family subsequently linked to a locus on chromosome 2q11, and it has since been reported in a
Jeroen H F de Baaij et al.
The Journal of biological chemistry, 287(17), 13644-13655 (2012-03-09)
Recently, mutations in the cyclin M2 (CNNM2) gene were identified to be causative for severe hypomagnesemia. In kidney, CNNM2 is a basolaterally expressed protein with predominant expression in the distal convoluted tubule. Transcellular magnesium (Mg(2+)) reabsorption in the distal convoluted
Bozena Polok et al.
American journal of human genetics, 84(2), 259-265 (2009-02-10)
Cone-rod dystrophies are inherited dystrophies of the retina characterized by the accumulation of deposits mainly localized to the cone-rich macular region of the eye. Dystrophy can be limited to the retina or be part of a syndrome. Unlike nonsyndromic cone-rod
Yosuke Funato et al.
The Journal of clinical investigation, 124(12), 5398-5410 (2014-10-28)
Intracellular Mg(2+) levels are strictly regulated; however, the biological importance of intracellular Mg(2+) levels and the pathways that regulate them remain poorly understood. Here, we determined that intracellular Mg(2+) is important in regulating both energy metabolism and tumor progression. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service