Skip to Content
Merck
All Photos(2)

Key Documents

HPA015788

Sigma-Aldrich

Anti-IKBKE antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-I kappa-B kinase epsilon, Anti-IKK- E, Anti-IKK-epsilon, Anti-IKK-i, Anti-IkBKE, Anti-Inducible I kappa-B kinase, Anti-Inhibitor of nuclear factor kappa-B kinase subunit epsilon

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,140.00

R$4,140.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,140.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,140.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

IHKLDKVNFSHLAKRLLQVFQEECVQKYQASLVTHGKRMRVVHETRNHLRLVGCSVAACNTEAQGVQESLSKLLEELSHQLLQDRAKGAQASPPPIAPYPSPTRKDLLLHMQELCEGMKLLASDLLDNNRIIERLN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IKBKE(9641)

Immunogen

Inhibitor of nuclear factor kappa-B kinase subunit epsilon recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Inhibitor of nuclear factor κ-B kinase subunit ε is an enzyme encoded by the IKBKE gene in humans. It interacts with NS2B/3 and acts as a novel target of DENV NS2B/3 protease. Knockdown of this gene may be associated with therapeutic advantages for HIV activation. It mainly functions during invasion and metastasis of ovarian cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73502

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yesseinia I Angleró-Rodríguez et al.
Clinical and vaccine immunology : CVI, 21(1), 29-38 (2013-11-01)
Dengue is the world's most common mosquito-borne viral infection and a leading cause of morbidity throughout the tropics and subtropics. Viruses are known to evade the establishment of an antiviral state by regulating the activation of interferon regulatory factor 3
Quanhong Sun et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 29(1), 90-102 (2013-06-25)
Paget's disease of bone (PDB) is characterized by abnormal osteoclasts with unique characteristics that include increased sensitivity of osteoclast progenitors to 1,25(OH)2 D3 , receptor activator of NF-κB ligand (RANKL), and TNF-α; increased osteoclast numbers; and increased expression of IL-6
Sarah Hsu et al.
Cancer research, 72(21), 5494-5504 (2012-09-04)
Inhibitor of IκB kinases (IKK) are key regulators of NF-κB signaling. Three IKK isoforms-α, β, and ε-have been linked to oncogenesis, yet the precise components of NF-κB signaling in ovarian cancer have not yet been dissected. We surveyed 120 ovarian
Guerau Fernandez et al.
Journal of virology, 87(7), 3966-3978 (2013-02-01)
Latently infected cell reservoirs represent the main barrier to HIV eradication. Combination antiretroviral therapy (cART) effectively blocks viral replication but cannot purge latent provirus. One approach to HIV eradication could include cART to block new infections plus an agent to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service