Skip to Content
Merck
All Photos(2)

Key Documents

HPA014459

Sigma-Aldrich

Anti-ST6GAL2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alpha 2,6-ST 2, Anti-Beta-galactoside alpha-2,6-sialyltransferase 2, Anti-CMP-N- acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2, Anti-ST6Gal II, Anti-ST6Gal-II, Anti-ST6GalII, Anti-Sialyltransferase 2, Anti-hST6Gal II

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Estimated to ship onMay 07, 2025



Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Estimated to ship onMay 07, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PSSLSFLETRRLLPVQGKQRAIMGAAHEPSPPGGLDARQALPRAHPAGSFHAGPGDLQKWAQSQDGFEHKEFFSSQVGRKSQSAFYPEDDDYFFAAGQPGWHSHTQGTLGFPSPGEPGPREGAFPAAQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ST6GAL2(84620)

General description

The gene ST6GAL2 (ST6 β-galactosamide α-2,6-sialyltranferase 2) belongs to the ST6Gal family that also includes another member, ST6Gal I. It is mainly expressed in embryonic and adult brain. It is also found to be expressed in small intestine and colon. The encoded protein is found to be 529 amino acids long and exhibits 48.9% amino acid sequence similarity with ST6Gal I. The ST6GAL2 gene is mapped to human chromosome 2q11.2-q12.1. It consists of eight exons spanning a length of 85kb. The protein contains type II transmembrane topology. All sialyltransferases of animal origin contain a short N-terminal tail that is cytoplasmic, a transmembrane domain, a stem region, and a catalytic domain with highly conserved motifs called sialylmotifs.

Immunogen

Beta-galactoside alpha-2,6-sialyltransferase 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene (ST6 β-galactosamide α-2,6-sialyltranferase 2) encodes a putative sialyltransferase. It catalyzes the transfer of sialic acid from CMP to an oligosaccharide substrate having Galβ1,4GlcNAc sequence at the nonreducing end of their carbohydrate groups through an α2, 6-linkage. It prefers the LacdiNAc structure as an acceptor substrate over the Galβ1-4GlcNAc structure.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72039

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ST6 β-Galactoside α-2, 6-Sialyltranferase 2 (ST6GAL2).
Takashima, S and Tsuji, S
Handbook of Glycosyltransferases and Related Genes?, 705-714 (2014)
Sylvain Lehoux et al.
Glycoconjugate journal, 27(1), 99-114 (2009-09-22)
The second human beta-galactoside alpha-2,6-sialyltransferase (hST6Gal II) differs from hST6Gal I, the first member of ST6Gal family, in substrate specificity and tissue expression pattern. While ST6GAL1 gene is expressed in almost all human tissues, ST6GAL2 shows a restricted tissue-specific pattern
Shou Takashima et al.
The Journal of biological chemistry, 277(48), 45719-45728 (2002-09-18)
A novel member of the human beta-galactoside alpha2,6-sialyltransferase (ST6Gal) family, designated ST6Gal II, was identified by BLAST analysis of expressed sequence tags and genomic sequences. The sequence of ST6Gal II encoded a protein of 529 amino acids, and it showed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service