Skip to Content
Merck
All Photos(5)

Key Documents

HPA012945

Sigma-Aldrich

Anti-ATP2B1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PMCA1, Anti-Plasma membrane calcium ATPase isoform 1, Anti-Plasma membrane calcium pump isoform 1, Anti-Plasma membrane calcium-transporting ATPase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

R$4,567.00


Estimated to ship onMay 28, 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

GDMANNSVAYSGVKNSLKEANHDGDFGITLAELRALMELRSTDALRKIQESYGDVYGICTKLKTSPNEGLSGNPADLERREAVFGKNFIP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP2B1(490)

General description

The gene encoding plasma membrane calcium-transporting ATPase 1 (ATP2B1) is located on chromosome 12q21-q23. It is localized to the basolateral membrane of cells and expressed widely in many tissues. ATP2B1 is known as the “housekeeping” isoform.

Immunogen

Plasma membrane calcium-transporting ATPase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Plasma membrane calcium-transporting ATPase 1 (ATP2B1) pumps Ca2+ out of the cell. It is thus involved in maintaining intracellular Ca2+ levels and regulates Ca2+ signaling in cells. ATP2B1 dysfunctioning has been shown to be involved with hypertension and arterial stiffness.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71544

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ján Lehotsky et al.
Frontiers in bioscience : a journal and virtual library, 7, d53-d84 (2002-01-10)
The biochemical function of the plasma membrane calcium ATPases (PMCAs) is the extrusion of cytosolic Ca2+ from the cell. Although this general function is well documented, the role of the complex isoform diversity and especially the contribution of specific isoforms
S Olson et al.
Genomics, 9(4), 629-641 (1991-04-01)
Human plasma membrane Ca2(+)-ATPase (PMCA) isoforms are encoded by at least four separate genes and the diversity of these enzymes is further increased by alternative splicing of transcripts. Cloned cDNAs for two of these isoforms have been used as probes
Qiuxin Wu et al.
Molecular and cellular biochemistry, 382(1-2), 263-272 (2013-07-03)
Ultraviolet B (UVB) could lead to the apoptosis of human lens epithelial cell and be hypothesized to be one of the important factors of cataractogenesis. In the human lens, Ca(2+)-ATPase is a major determinant of calcium homeostasis. Plasma membrane calcium
Michael C Chicka et al.
The Journal of biological chemistry, 278(20), 18464-18470 (2003-03-08)
Plasma membrane Ca(2+)-ATPases (PMCAs) are involved in local Ca(2+) signaling and in the spatial control of Ca(2+) extrusion, but how different PMCA isoforms are targeted to specific membrane domains is unknown. In polarized MDCK epithelial cells, a green fluorescent protein-tagged
Geoffrey M Goellner et al.
Annals of the New York Academy of Sciences, 986, 461-471 (2003-05-24)
Plasma membrane Ca(2+) ATPases (PMCAs) maintain intracellular Ca(2+) homeostasis and participate in the local regulation of Ca(2+) signaling. Spatially separate demands for Ca(2+) regulation require proper membrane targeting of PMCAs, but the mechanism of PMCA targeting is unknown. Using the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service