Skip to Content
Merck
All Photos(5)

Key Documents

HPA012825

Sigma-Aldrich

Anti-FNDC3A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fibronectin type-III domain-containing protein 3a antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

VNWEVPLSNGTDVTEYRLEWGGVEGSMQICYCGPGLSYEIKGLSPATTYYCRVQALSVVGAGPFSEVVACVTPPSVPGIVTCLQEISDDEIENPHYSPSTCLAISWEKPCDHGSEILAYSIDFGDKQSLTVGKV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FNDC3A(22862)

Looking for similar products? Visit Product Comparison Guide

General description

Fibronectin type-III domain-containing protein 3A (FNDC3A) contains a proline-rich region at the N-terminus, eight type III-fibronectin modules and a trans membrane helix at the C-terminus. It is present in the Golgi vesicles. FNDC3A is expressed in odontoblasts, in nerve fibers of the dental pulp and tissues of the trachea, brain and kidney. The gene encoding this protein is located on chromosome 13q14.

Immunogen

Fibronectin type-III domain-containing protein 3a recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Single nucleotide polymorphism in fibronectin type-III domain-containing protein 3A (FNDC3A) has been shown to be associated with autism spectrum disorder (ASD). In mice, FNDC3A mediates spermatid–sertoli adhesion during spermatogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71726

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alain Latil et al.
The Prostate, 57(1), 39-50 (2003-07-30)
Loss of heterozygosity (LOH) on chromosome arm 13q14 is one of the most consistent genetic alterations in sporadic prostate cancer. This alteration may be involved in prostate oncogenesis through inactivation of one or more tumor suppressor genes (TSGs). Candidate gene
Myungja Ro et al.
Psychiatry research, 209(2), 246-248 (2013-05-04)
Autism spectrum disorder (ASD) is a neurodevelopmental syndrome associated with impairments of reciprocal communication and cognitive function. Associations between single-nucleotide polymorphisms (SNPs) and ASD were analysed by logistic regression. Polymorphisms in fibronectin type III domain-containing 3A (FNDC3A) exhibited significant associations
Kevin L Obholz et al.
Developmental biology, 298(2), 498-513 (2006-08-15)
Symplastic spermatids (sys) male mice are sterile due to a recessive mutation that causes defective adhesion between spermatids and Sertoli cells within the seminiferous epithelium. We show that the mutation in sys mice involves a deletion of 1.24 Mb of
F Carrouel et al.
Journal of dental research, 87(2), 131-136 (2008-01-26)
Previously, we established a subtractive cDNA library enriched in odontoblast-specific genes and hypothesized that new, previously unidentified, markers would be present, associated with the odontoblast phenotype. In this paper, we report the first characterization of a new gene we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service