Skip to Content
Merck
All Photos(2)

Key Documents

HPA010819

Sigma-Aldrich

Anti-BAMBI antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

Synonym(s):

NMA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

NKRLQDQRQQMLSRLHYSFHGHHSKKGQVAKLDLECMVPVSGHENCCLTCDKMRQADLSNDKILSLVHWGMYSGHGKLEFV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAMBI(25805)

General description

Bone morphogenic protein and activin membrane-bound inhibitor (BAMBI) is a transmembrane glycoprotein which is a pseudo receptor belonging to the family of TGF-β (transforming growth factor beta receptor) type I receptors. BAMBI is predominantly expressed in placenta, spleen and kidney medulla and also found liver, prostate, gut and kidney cortex.

Immunogen

BMP and activin membrane-bound inhibitor

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Bone morphogenic protein and activin membrane-bound inhibitor (BAMBI) acts as an inhibitor of TGF-β (transforming growth factor beta receptor) signaling which causes epithelial-mesenchymal transition. Upregulation of BAMBI gene is observed in chronic obstructive pulmonary disease and a number of cancers including thyroid, liver, colorectal cancer and ovarian cancer. BAMBI might regulate the events of inflammation and remodeling processes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71703

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dysregulation of the Transforming Growth Factor β Pathway in Induced Pluripotent Stem Cells Generated from Patients with Diamond Blackfan Anemia
Jingping G, et al.
PLoS ONE, 10(8), e0134878-e0134878 (2015)
Downregulation of the TGF? Pseudoreceptor BAMBI in Non?Small Cell Lung Cancer Enhances TGF? Signaling and Invasion
Marwitz S, et al.
Cancer Research (2016)
LPS/TLR4 Signaling Enhances TGF-β Response Through Downregulating BAMBI During Prostatic Hyperplasia
Yao H, et al.
Scientific Reports, 6, 27051-27051 (2016)
Kai Liu et al.
Molecular medicine reports, 10(2), 874-880 (2014-06-11)
The upregulation of bone morphogenetic protein and activin membrane‑bound inhibitor (BAMBI) has been observed in several types of malignant cancer, including thyroid, ovarian, liver and colorectal cancer. However, the pathological role and the regulatory mechanism of BAMBI in gastric cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service