Skip to Content
Merck
All Photos(7)

Key Documents

HPA010010

Sigma-Aldrich

Anti-RPS6KB2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-70 kDa ribosomal protein S6 kinase 2, Anti-Ribosomal protein S6 kinase beta-2, Anti-S6 kinase-related kinase, Anti-S6K-beta, Anti-S6K2, Anti-SRK, Anti-p70 S6 kinase beta, Anti-p70 S6Kbeta, Anti-p70 ribosomal S6 kinase beta, Anti-p70-S6KB, Anti-p70-beta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,140.00

R$4,140.00


Estimated to ship onApril 23, 2025



Select a Size

Change View
100 μL
R$4,140.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,140.00


Estimated to ship onApril 23, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

VDSPDDTALSESANQAFLGFTYVAPSVLDSIKEGFSFQPKLRSPRRLNSSPRAPVSPLKFSPFEGFRPSPSLPEPTELPLPPLLPPPPPSTTAPLPIRPPSGTKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPS6KB2(6199)

General description

RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2), also called 40S ribosomal protein S6 kinase (S6K2), and S6k1 form the two homologs of S6K protein. It is a serine/threonine kinase, which was first recognized by its homology to S6K1.

Immunogen

Ribosomal protein S6 kinase beta-2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2) is a cellular protein, which is a downstream target of mTOR (mammalian target of rapamycin). It also phosphorylates S6 protein. The function of RPS6KB2 is not fully understood yet. In small cell lung cancer, this protein is thought to be essential for chemoresistance mediated by FGF2 (fibroblast growth factor 2). It also controls the activity of Akt/ protein kinase B (PKB) and cell survival. This protein is constitutively expressed during cell cycle, with peak levels during G and M phase, suggesting it might play important roles in both phases. It crosstalks with hormone receptor signaling and is involved in breast carcinogenesis, and thus, might have potential as marker for prognosis and hormone therapy response in breast cancer. RPS6KB2 is also amplified in gastric cancer and is a marker of poor prognosis of the same.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70180

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elin Karlsson et al.
Breast cancer research : BCR, 15(5), R96-R96 (2013-10-18)
mTOR and its downstream effectors the 4E-binding protein 1 (4EBP1) and the p70 ribosomal S6 kinases (S6K1 and S6K2) are frequently upregulated in breast cancer, and assumed to be driving forces in tumourigenesis, in close connection with oestrogen receptor (ER)
Shuhei Yoshida et al.
Anticancer research, 33(2), 469-475 (2013-02-09)
The gene amplification of ribosomal protein S6 kinase 1 and 2 (S6K1 and S6K2) and its clinical relevance in gastric cancer remain unclear. A comparative genomic hybridization analysis and DNA copy number assay were performed for nine cancer cell lines.
Derek Boyer et al.
Molecular and cellular biochemistry, 307(1-2), 59-64 (2007-09-06)
Ribosomal S6 kinase 2 (S6K2) is one of the kinases regulated by the mammalian target of rapamycin (mTOR) signaling pathway. Although it has been identified as a kinase homologous to S6K1, evidence suggests that the two kinases have non-overlapping functions
Savitha Sridharan et al.
Cancer research, 71(7), 2590-2599 (2011-03-24)
The 40S ribosomal protein S6 kinase (S6K) acts downstream of mTOR, which plays important roles in cell proliferation, protein translation, and cell survival and is a target for cancer therapy. mTOR inhibitors are, however, of limited success. Although Akt is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service