Skip to Content
Merck
All Photos(4)

Key Documents

HPA008732

Sigma-Aldrich

Anti-MUTYH antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A/G-specific adenine DNA glycosylase, Anti-MutY homolog, Anti-hMYH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,140.00

R$4,140.00


Estimated to ship onJune 01, 2025



Select a Size

Change View
100 μL
R$4,140.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,140.00


Estimated to ship onJune 01, 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PDVEECAPNTGQCHLCLPPSEPWDQTLGVVNFPRKASRKPPREESSATCVLEQPGALGAQILLVQRPNSGLLAGLWEFPSVTWEPSEQLQRKALLQELQRWA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUTYH(4595)

General description

MUTYH (mutY DNA glycosylase) is a key enzyme of the human DNA repair system. It is expressed in several isoforms, of which two are predominant- type1 mitochondrial and type2 nuclear form.[1] This gene is localized to human chromosome 1p34.1 and spans 11.2kb.

Immunogen

A/G-specific adenine DNA glycosylase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MUTYH (mutY DNA glycosylase) is a part of the DNA glycosylase-initiating base excision repair, where it catalyzes the excision of mispaired adenine with 8-Hydroxyguanine (8OHG) is double-stranded DNA. Biallelic germline inactivating mutations in this gene are responsible for MUTYH-associated polyposis, which results in multiple colorectal polyps and carcinoma. Studies in a Japanese patient show that p.Arg109Trp variant of this gene results in dysfunctional MUTYH enzyme. This results in early-onset colorectal cancer.[1] Inactivation of this gene leads to genetic instability in lymphoblastoid cell lines, which is dependent on the degree of inactivation. Biallelic mutations in this gene are also responsible for Lynch-like syndrome (LLS), which is characterized by tumors containing mismatch repair (MMR) deficiency, without association with germline mutations or somatic methylation of MMR genes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71064

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuya Shinmura et al.
Oxidative medicine and cellular longevity, 2014, 617351-617351 (2014-05-07)
The biallelic inactivation of the 8-hydroxyguanine repair gene MUTYH leads to MUTYH-associated polyposis (MAP), which is characterized by colorectal multiple polyps and carcinoma(s). However, only limited information regarding MAP in the Japanese population is presently available. Since early-onset colorectal cancer
Francesca Grasso et al.
Human molecular genetics, 23(14), 3843-3852 (2014-02-27)
The MUTYH DNA glycosylase counteracts mutagenesis by removing adenine misincorporated opposite DNA 8-oxo-7,8-dihydro-2'-deoxyguanosine (8-oxodG). Biallelic germline mutations in MUTYH cause the autosomal recessive MUTYH-associated polyposis (MAP). The impact on genetic instability of the p.Tyr179Cys and p.Arg245His MUTYH variants was evaluated
Adela Castillejo et al.
European journal of cancer (Oxford, England : 1990), 50(13), 2241-2250 (2014-06-24)
Individuals with tumours showing mismatch repair (MMR) deficiency not linked to germline mutations or somatic methylation of MMR genes have been recently referred as having 'Lynch-like syndrome' (LLS). The genetic basis of these LLS cases is unknown. MUTYH-associated polyposis patients
Giovana T Torrezan et al.
BMC medical genetics, 12, 128-128 (2011-10-04)
MUTYH-associated polyposis (MAP) is a recessive, hereditary, colorectal cancer-predisposing syndrome caused by biallelic mutations in the MUTYH gene. Most MUTYH pathogenic variants are missense mutations, and until recently no gross genomic deletions had been described. We have identified a large

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service