Skip to Content
Merck
All Photos(3)

Key Documents

HPA007342

Sigma-Aldrich

Anti-TCTA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-T-cell leukemia translocation-altered gene protein, Anti-T-cell leukemia translocation-associated gene protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Estimated to ship onApril 20, 2025



Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Estimated to ship onApril 20, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AESWSGQALQALPATVLGALGSEFLREWEAQDMRVTLFKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TCTA(6988)

General description

TCTA (T-cell leukemia translocation altered) gene is localized at the site of a t(1;3)(p34;p21) translocation breakpoint in T-cell acute lymphoblastic leukemia. The gene is highly conserved from Drosophila to humans. It is ubiquitously expressed in normal tissues and most abundantly expressed in the kidney.

Immunogen

T-cell leukemia translocation-altered gene protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TCTA (T-cell leukemia translocation altered) protein may function in cellular fusion during osteoclastogenesis. It also has a role in human tumorigenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70156

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P D Aplan et al.
Cancer research, 55(9), 1917-1921 (1995-05-01)
We have cloned and characterized a novel gene at the site of a t(1;3)(p34;p21) translocation breakpoint in T-cell acute lymphoblastic leukemia. A cDNA for this gene, for which we propose the designation TCTA (T-cell leukemia translocation-associated gene), has been cloned.
Shigeru Kotake et al.
Journal of biomedicine & biotechnology, 2012, 675317-675317 (2011-12-17)
Synovial tissues of patients with rheumatoid arthritis (RA) include factors regulating bone resorption, such as receptor activator NF-κB ligand (RANKL), TNF-α, IL-6, IL-17, and IFN-γ. However, in addition to these cytokines, other factors expressed in synovial tissues may play a
Shigeru Kotake et al.
Bone, 45(4), 627-639 (2009-06-30)
Synovial tissues of patients with rheumatoid arthritis (RA) include factors regulating bone resorption, such as receptor activator NF-kappaB ligand (RANKL), TNFalpha, IL-6, IL-17 and IFNgamma. However, in addition to these cytokines, other factors expressed in synovial tissues may play a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service