Skip to Content
Merck
All Photos(3)

Key Documents

HPA007053

Sigma-Aldrich

ANTI-CDK19 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDC2- related protein kinase 6, Anti-CDC2L6, Anti-Cell division cycle 2-like protein kinase 6, Anti-Cyclin-dependent kinase 11, Anti-Death-preventing kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.4 μg/mL
immunofluorescence: 1-4 μg/mL

immunogen sequence

KGDKNQQQQQNQHQQPTAPPQQAAAPPQAPPPQQNSTQTNGTAGGAGAGVGGTGAGLQHSQDSSLNQVPPNKKPRLGPSGANSGGPVMPSDYQHSSSRLNYQSSVQGSSQSQSTLGYSSSSQQSSQYHPSH

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CDC2L6(23097)

Immunogen

Cell division cycle 2-like protein kinase 6 recombinant protein epitope signature tag (PrEST)

Application

Anti-CDC2L6 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CDK19 (cyclin-dependent kinase 19) is a cyclin-dependent kinase protein. In transcription, it acts as a negative transcriptional regulator. It functions as a subunit of human CDK8-independent mediator complex in the transcriptional activation by RNA polymerase II. In conjugated form with CDK8, it exerts CTD kinase activity for the serine residues.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71438.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Brägelmann et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(7), 1829-1840 (2016-09-30)
Purpose: The Mediator complex is a multiprotein assembly, which serves as a hub for diverse signaling pathways to regulate gene expression. Because gene expression is frequently altered in cancer, a systematic understanding of the Mediator complex in malignancies could foster
Finn Becker et al.
International journal of cancer, 146(2), 577-588 (2019-07-05)
The Mediator complex is a transcriptional regulator interacting with transcription factors and RNA-polymerase-II. Recently, we identified its subunit CDK19 to be specifically expressed in prostate cancer (PCa) and to be functionally implicated in PCa aggressiveness. Aim of our study was
Taiki Tsutsui et al.
Genes to cells : devoted to molecular & cellular mechanisms, 16(12), 1208-1218 (2011-11-29)
Mediator is a large complex containing up to 30 subunits that consist of four modules each: head, middle, tail and CDK/Cyclin. Recent studies have shown that CDK8, a subunit of the CDK/Cyclin module, is one of the key subunits of
Agnieszka Witalisz-Siepracka et al.
Cancer immunology research, 6(4), 458-466 (2018-02-02)
Cyclin-dependent kinase 8 (CDK8) is a member of the transcription-regulating CDK family. CDK8 activates or represses transcription by associating with the mediator complex or by regulating transcription factors. Oncogenic activity of CDK8 has been demonstrated in several cancer types. Targeting
Min Ju Park et al.
The Journal of biological chemistry, 293(13), 4870-4882 (2018-02-15)
Somatic mutations in exon 2 of the RNA polymerase II transcriptional Mediator subunit MED12 occur at high frequency in uterine fibroids (UFs) and breast fibroepithelial tumors as well as recurrently, albeit less frequently, in malignant uterine leimyosarcomas, chronic lymphocytic leukemias

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service