Skip to Content
Merck
All Photos(3)

Key Documents

HPA003231

Sigma-Aldrich

Anti-CHM antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Choroideraemia protein antibody produced in rabbit, Anti-REP-1 antibody produced in rabbit, Anti-Rab escort protein 1 antibody produced in rabbit, Anti-Rab proteins geranylgeranyltransferase component A 1 antibody produced in rabbit, Anti-TCD protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

SRKDKTIQHVEVFCYASQDLHEDVEEAGALQKNHALVTSANSTEAADSAFLPTEDESLSTMSCEMLTEQTPSSDPENALEVNGAEVTGEKENHCDDKTCVPSTSAEDMSENVPIAEDTTEQPKKNRITYSQIIKEG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHM(1121)

General description

The gene CHM (choroideremia) is mapped to human chromosome Xq21.2. It encodes a 653 amino acid protein. It contains 15 exons spanning a length of 150kb.

Immunogen

Rab proteins geranylgeranyltransferase component A 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-CHM antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CHM (choroideremia) gene encodes a protein that is also called as Rab proteins geranylgeranyltransferase component A 1. This subunit is a substrate-binding subunit of the Rab geranylgeranyl transferase (RGGT) holoenzyme. It binds unprenylated Rab proteins and presents them to the catalytic component B of Rab-GGTase. Alternatively, the subunit encoded by this gene first binds to RGGT, and this complex then binds and prenylates Rab proteins. These Rab proteins are small GTPases that function in regulating vesicular membrane traffic. Mutations in this gene can cause choroideremia, also referred to as tapetochoroidal dystrophy (TCD), that is characterized by progressive dystrophy of the choroid, retinal pigment epithelium and retina.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74342

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanmin Xue et al.
Nature medicine, 24(10), 1507-1512 (2018-10-10)
Retinal gene therapy is increasingly recognized as a novel molecular intervention that has huge potential in treating common causes of blindness, the majority of which have a genetic aetiology1-5. Choroideremia is a chronic X-linked retinal degeneration that was first described
K Alexandrov et al.
The EMBO journal, 13(22), 5262-5273 (1994-11-15)
Rab proteins comprise a family of small GTPases that serve a regulatory role in vesicular membrane traffic. Geranylgeranylation of these proteins on C-terminal cysteine motifs is crucial for their membrane association and function. This post-translational modification is catalysed by rab
Rudi A Baron et al.
The Biochemical journal, 415(1), 67-75 (2008-06-06)
Prenylation (or geranylgeranylation) of Rab GTPases is catalysed by RGGT (Rab geranylgeranyl transferase) and requires REP (Rab escort protein). In the classical pathway, REP associates first with unprenylated Rab, which is then prenylated by RGGT. In the alternative pathway, REP
J A van den Hurk et al.
Human mutation, 9(2), 110-117 (1997-01-01)
Choroideremia (CHM) is an X-linked recessive eye disease that results from mutations involving the Rab escort protein-1 (REP-1) gene. In 18 patients deletions of different sizes have been found. Two females suffering from CHM were reported to have translocations that
Aaron Black et al.
The journal of gene medicine, 16(5-6), 122-130 (2014-06-26)
Choroideremia (CHM) is a slowly progressive X-linked retinal degeneration that results in a loss of photoreceptors, retinal pigment epithelium and choroid. CHM, the gene implicated in choroideremia, encodes Rab escort protein-1 (REP-1), which is involved in the post-translational activation via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service