Skip to Content
Merck
All Photos(3)

Key Documents

HPA002697

Sigma-Aldrich

Anti-SSRP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Chromatin- specific transcription elongation factor 80 kDa subunit, Anti-FACT 80 kDa subunit, Anti-FACT complex subunit SSRP1, Anti-FACTp80, Anti-Facilitates chromatin transcription complex subunit SSRP1, Anti-Structure-specific recognition protein 1, Anti-hSSRP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,140.00

R$4,140.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,140.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,140.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DDAEVSLMEVRFYVPPTQEDGVDPVEAFAQNVLSKADVIQATGDAICIFRELQCLTPRGRYDIRIYPTFLHLHGKTFDYKIPYTTVLRLFLLPHKDQRQMFFVISLDPPIKQGQTRYHFLILLFSKDEDISLTLNMNEEEVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SSRP1(6749)

General description

Structure specific recognition protein 1 (SSRP1), a nuclear protein, is involved in the regulation of mitotic machinery of microtubules (MTs) in cell cycle progression. It is present with the astral and spindle MTs. It is upregulated in proliferating tissues in the mouse and human cancerous tissues. It is also present in less-renewable and differentiated tissues or cells.

Immunogen

FACT complex subunit SSRP1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Structure specific recognition protein 1 (SSRP1) is associated with the replication-associated repair processes. During transcription elongation, it acts as a subunit of FACT (facilitates chromatin transcription) complex. FACT, a histone chaperone, is essential for chromatin recovery or remodeling during transcription and cell differentiation. It also phosphorylates the human p53 protein at Ser-392 position in association with CK2 (casein kinase II). SSRP1 also acts as a co-activator of a transcriptional activator, p63, to enhance the p63γ-dependent luciferase activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86222

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shelya X Zeng et al.
The EMBO journal, 21(20), 5487-5497 (2002-10-11)
The p53 homolog p63 is a transcriptional activator. Here, we describe the identification of an HMG1-like protein SSRP1 as a co-activator of p63. Over expression of wild-type, but not deletion mutant, SSRP1 remarkably enhanced p63gamma-dependent luciferase activity, G1 arrest, apoptosis
Shelya X Zeng et al.
Molecular and cellular biology, 30(4), 935-947 (2009-12-10)
Tight regulation of microtubule (MT) dynamics is essential for proper chromosome movement during mitosis. Here we show, using mammalian cells, that structure-specific recognition protein 1 (SSRP1) is a novel regulator of MT dynamics. SSRP1 colocalizes with the spindle and midbody
Fu-Kai Hsieh et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(19), 7654-7659 (2013-04-24)
FACT (facilitates chromatin transcription) is a histone chaperone that promotes chromatin recovery during transcription, with additional roles in cell differentiation. Although several models of the action of FACT during transcription have been proposed, they remain to be experimentally evaluated. Here
G Orphanides et al.
Cell, 92(1), 105-116 (1998-03-07)
The requirements for transcriptional activation by RNA polymerase II were examined using chromatin templates assembled in vitro and a transcription system composed of the human general transcription factors and RNA polymerase II. Activator-induced, energy-dependent chromatin remodeling promoted efficient preinitiation complex
Anuradha Kumari et al.
Journal of cellular biochemistry, 108(2), 508-518 (2009-07-30)
A possible role for structure-specific recognition protein 1 (SSRP1) in replication-associated repair processes has previously been suggested based on its interaction with several DNA repair factors and the replication defects observed in SSRP1 mutants. In this study, we investigated the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service