Skip to Content
Merck
All Photos(6)

Key Documents

HPA001794

Sigma-Aldrich

Anti-ZNF268 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HZF3, Anti-zinc finger protein 268

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Estimated to ship onApril 06, 2025



Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Estimated to ship onApril 06, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AQVPNQTCPNTVWKIDDLMDWHQENKDKLGSTAKSFECTTFGKLCLLSTKYLSRQKPHKCGTHGKSLKYIDFTSDYARNNPNGFQVHGKSFFHSKHEQTVIGIKYCESIESGKTVNKKSQL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF268(10795)

Immunogen

zinc finger protein 268 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZNF268 (zinc finger protein 268) is a krüppel-associated box (KRAB)-containing, C2H2 zinc finger (KRAB-ZNF) protein expressed in human fetal tissues and tumor derived cell lines. It is more predominantly expressed in early embryonic stage. The zinc finger domain binds to the target DNA sequences to control gene expression, followed by KRAB induced transcription repression. It has been reported that activity of ZNF268 is involved with the human foetal liver development, haematological diseases and cervical cancer development. It is also associated with the differentiation of blood cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83487

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Hu et al.
Oncology letters, 6(1), 49-54 (2013-08-16)
Ovarian cancer is one of the most lethal gynaecological cancers worldwide. However, the mechanisms underlying ovarian carcinogenesis are not well understood. The present study used immunostaining, western blotting and quantitative real-time PCR to demonstrate that ZNF268 is overexpressed in human
Huanjie Shao et al.
International journal of molecular medicine, 18(3), 457-463 (2006-07-26)
The ZNF268 gene was originally isolated from an early human embryo cDNA library. Several different transcripts have been isolated for the ZNF268 gene and developmental expression studies suggest that ZNF268 plays a role in the development of human fetal liver
Ming-Xiong Guo et al.
The Journal of biological chemistry, 281(34), 24623-24636 (2006-06-22)
Human ZNF268 gene is a typical Krüppel-associated box/C2H2 zinc finger gene whose homolog has been found only in higher mammals and not in lower mammals such as mouse. Its expression profiles have suggested that it plays a role in the
Yan Sun et al.
International journal of molecular medicine, 14(6), 971-975 (2004-11-18)
The advancement in gene knockout and transgenesis have brought about enormous improvement in our understanding of mouse embryogenesis in the past decade or so. On the other hand, relatively little is known about human embryogenesis due largely to the lack

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service