Skip to Content
Merck
All Photos(3)

Key Documents

HPA001202

Sigma-Aldrich

Anti-RNF139 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-RING finger protein 139 antibody produced in rabbit, Anti-Translocation in renal carcinoma on chromosome 8 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

R$4,567.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PEIKGSRLQEINDVCAICYHEFTTSARITPCNHYFHALCLRKWLYIQDTCPMCHQKVYIEDDIKDNSNVSNNNGFIPPNETPEEAVREAAAESDRELNEDDSTDCDDDVQRERNGVIQHTGAAAEEF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF139(11236)

Immunogen

RING finger protein 139 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RNF139 (ring finger protein 139) gene encodes a endoplasmic reticulum protein that contains a RING-H2 finger required for Ub ligase activity and a sterol-sensing domain. It possesses E3-ubiquitin ligase activity. It negatively regulates cell proliferation via mechanisms involving G2/M arrest, decreased DNA synthesis and cell death. It acts as a putative tumor suppressor. It represses genes involved in the biosynthesis of cholesterol and fatty acids that are transcriptionally regulated by the sterol response element binding proteins (SREBPs), thus, establishing a link between growth control and the cholesterol/lipid homeostasis pathway. It inhibits the transport of sterol regulatory element-binding protein-2 (SREBP-2) from ER to Golgi. It is linked to the development of hereditary kidney cancer. The protein is required for the dislocation of MHC I (major histocompatibility complex class I) from the ER.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79893

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Brauweiler et al.
Oncogene, 26(16), 2263-2271 (2006-10-04)
TRC8/RNF139 and von Hippel-Lindau (VHL) both encode E3 ubiquitin (Ub) ligases mutated in clear-cell renal carcinomas (ccRCC). VHL, inactivated in nearly 70% of ccRCCs, is a tumor suppressor encoding the targeting subunit for a Ub ligase complex that downregulates hypoxia-inducible
Robert M Gemmill et al.
Oncogene, 21(22), 3507-3516 (2002-05-29)
VHL is part of an SCF related E3-ubiquitin ligase complex with 'gatekeeper' function in renal carcinoma. However, no mutations have been identified in VHL interacting proteins in wild type VHL tumors. We previously reported that the TRC8 gene was interrupted
Masato Irisawa et al.
The Journal of biological chemistry, 284(42), 28995-29004 (2009-08-27)
TRC8 (translocation in renal cancer from chromosome 8) is an intrinsic protein of the endoplasmic reticulum that contains a sterol-sensing domain and a RING finger motif encoding an E3 ubiquitin ligase. Here we show that TRC8 overexpression hinders sterol regulatory
Helen R Stagg et al.
The Journal of cell biology, 186(5), 685-692 (2009-09-02)
The US2 and US11 gene products of human cytomegalovirus promote viral evasion by hijacking the endoplasmic reticulum (ER)-associated degradation (ERAD) pathway. US2 and US11 initiate dislocation of newly translocated major histocompatibility complex class I (MHC I) from the ER to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service