Skip to Content
Merck
All Photos(2)

Key Documents

HPA000911

Sigma-Aldrich

Anti-FGD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FYVE, RhoGEF and PH domain-containing protein 1 antibody produced in rabbit, Anti-Faciogenital dysplasia 1 protein antibody produced in rabbit, Anti-Rho/Rac GEF antibody produced in rabbit, Anti-Rho/Rac guanine nucleotide exchange factor FGD1 antibody produced in rabbit, Anti-Zinc finger FYVE domain-containing protein 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$4,567.00

R$4,567.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$4,567.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

R$4,567.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

western blot: 0.04-0.4 μg/mL

immunogen sequence

DSDPGASEPGLLARRGSGSALGGPLDPQFVGPSDTSLGAAPGHRVLPCGPSPQHHRALRFSYHLEGSQPRPGLHQGNRILVKSLSLDPGQSLEPHPEGPQRLRSDP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGD1(2245)

Immunogen

FYVE, RhoGEF and PH domain-containing protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FYVE, RhoGEF and PH domain-containing protein 1 (FGD1) is a protein encoded by the FGD1 gene in humans. It is mapped to chromosome Xp11.21. It encodes a guanine nucleotide exchange factor, specifically activating Rho GTPase cell division cycle 42 (Cdc42). Dysfunction of the gene causes Aarskog-Scott syndrome, which is an X-linked disorder that may affect bone and intellectual development. The Aarskog-Scott syndrome, also known as Faciogenital dysplasia (FGDY), is characterized by disproportionately short stature and by facial, skeletal and urogenital anomalies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74066

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Li et al.
Genetics and molecular research : GMR, 13(1), 127-133 (2014-01-22)
FGD1 encoding a guanine nucleotide exchange factor, specifically activates Rho GTPase cell division cycle 42 (Cdc42). Dysfunction of FGD1 causes Aarskog-Scott syndrome (MIM #305400), an X-linked disorder that may affect bone and intellectual development. However, the relationship between FGD1 and
N G Pasteris et al.
Cell, 79(4), 669-678 (1994-11-18)
Faciogenital dysplasia (FGDY), also known as Aarskog-Scott syndrome, is an X-linked developmental disorder characterized by disproportionately short stature and by facial, skeletal, and urogenital anomalies. Molecular genetic analyses mapped FGDY to chromosome Xp11.21. To clone this gene, YAC clones spanning
Sylvie Thuault et al.
Scientific reports, 10(1), 6787-6787 (2020-04-24)
Metastatic progression is the leading cause of mortality in breast cancer. Invasive tumor cells develop invadopodia to travel through basement membranes and the interstitial matrix. Substantial efforts have been made to characterize invadopodia molecular composition. However, their full molecular identity
Thomas Daubon et al.
Journal of cell science, 129(13), 2586-2598 (2016-05-28)
Podosomes are dynamic cell-matrix contact structures that combine several key abilities, including adhesion, matrix degradation and mechanosensing. These actin-based cytoskeletal structures have been mostly studied in monocytic cells, but much less is known about those formed in other lineages. In
Dan Georgess et al.
European journal of cell biology, 97(8), 533-545 (2018-10-06)
Actin subunits assemble into actin filaments whose dynamics and three-dimensional architectures are further regulated by a variety of cellular factors to establish the functional actin cytoskeleton. The C-glucosidic ellagitannin vescalagin and its simpler analogue vescalin, affect both the dynamics and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service